Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132589189-132589448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARHL1
Entrez Gene ID
56751
Gene Name
BarH like homeobox 1
Gene Aliases
-
UniGene
Hs.283809
Gene Chromosome Location
Chr.9: 132582606-132590253 on Build GRCh38
Transcripts
NM_020064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1314587 c.806C>G p.S269C Chr.9: 132589344-132589344 on Build GRCh38
6044311 c.889C>A p.R297S Chr.9: 132589427-132589427 on Build GRCh38
607886 c.715G>A p.V239I Chr.9: 132589253-132589253 on Build GRCh38
3905114 c.792G>A p.P264P Chr.9: 132589330-132589330 on Build GRCh38
3905115 c.825G>A p.A275A Chr.9: 132589363-132589363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765313556 Chr.9: 132589400-132589401 on Build GRCh38
rs769270458 Chr.9: 132589333-132589333 on Build GRCh38
rs923993915 Chr.9: 132589295-132589295 on Build GRCh38
rs922543244 Chr.9: 132589312-132589312 on Build GRCh38
rs750641828 Chr.9: 132589270-132589270 on Build GRCh38
rs763315869 Chr.9: 132589235-132589235 on Build GRCh38
rs777831471 Chr.9: 132589299-132589299 on Build GRCh38
rs751536490 Chr.9: 132589384-132589384 on Build GRCh38
rs770183070 Chr.9: 132589231-132589231 on Build GRCh38
rs200018434 Chr.9: 132589267-132589267 on Build GRCh38
rs775662670 Chr.9: 132589233-132589233 on Build GRCh38
rs758244911 Chr.9: 132589373-132589373 on Build GRCh38
rs985247553 Chr.9: 132589356-132589356 on Build GRCh38
rs766626847 Chr.9: 132589282-132589282 on Build GRCh38
rs111570450 Chr.9: 132589255-132589255 on Build GRCh38
rs200567501 Chr.9: 132589405-132589405 on Build GRCh38
rs752499320 Chr.9: 132589288-132589288 on Build GRCh38
rs764549827 Chr.9: 132589241-132589241 on Build GRCh38
rs764037500 Chr.9: 132589379-132589379 on Build GRCh38
rs756789300 Chr.9: 132589318-132589318 on Build GRCh38
rs759791432 Chr.9: 132589392-132589394 on Build GRCh38
rs762296297 Chr.9: 132589350-132589350 on Build GRCh38
rs377171961 Chr.9: 132589428-132589428 on Build GRCh38
rs749851046 Chr.9: 132589416-132589416 on Build GRCh38
rs766471953 Chr.9: 132589360-132589360 on Build GRCh38
rs746984017 Chr.9: 132589302-132589302 on Build GRCh38
rs754152922 Chr.9: 132589364-132589364 on Build GRCh38
rs756231019 Chr.9: 132589273-132589273 on Build GRCh38
rs757072853 Chr.9: 132589396-132589396 on Build GRCh38
rs769061368 Chr.9: 132589347-132589347 on Build GRCh38
rs776316564 Chr.9: 132589219-132589219 on Build GRCh38
rs761044647 Chr.9: 132589311-132589311 on Build GRCh38
rs745575701 Chr.9: 132589227-132589227 on Build GRCh38
rs779662889 Chr.9: 132589427-132589427 on Build GRCh38
rs749610036 Chr.9: 132589341-132589341 on Build GRCh38
rs774884359 Chr.9: 132589246-132589246 on Build GRCh38
rs781200412 Chr.9: 132589408-132589408 on Build GRCh38
rs770397094 Chr.9: 132589207-132589207 on Build GRCh38
rs1014917806 Chr.9: 132589224-132589224 on Build GRCh38
rs925812608 Chr.9: 132589389-132589389 on Build GRCh38
rs775142120 Chr.9: 132589340-132589340 on Build GRCh38
rs201426274 Chr.9: 132589348-132589348 on Build GRCh38
rs760832863 Chr.9: 132589358-132589358 on Build GRCh38
rs761847169 Chr.9: 132589261-132589261 on Build GRCh38
rs773109875 Chr.9: 132589357-132589357 on Build GRCh38
rs745386325 Chr.9: 132589331-132589331 on Build GRCh38
rs549093104 Chr.9: 132589294-132589294 on Build GRCh38
rs780627128 Chr.9: 132589321-132589321 on Build GRCh38
rs755677545 Chr.9: 132589421-132589421 on Build GRCh38

View Full Product Details