Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 8791124-8791373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9A
Entrez Gene ID
171482
Gene Name
family with sequence similarity 9 member A
Gene Aliases
TEX39A
UniGene
Hs.382062
Gene Chromosome Location
Chr.X: 8790795-8801383 on Build GRCh38
Transcripts
NM_001171186.1 NM_174951.3

SNP Detail

SNP ID Chromosome Location
rs374401317 Chr.X: 8791339-8791339 on Build GRCh38
rs1018333040 Chr.X: 8791180-8791180 on Build GRCh38
rs61732570 Chr.X: 8791291-8791291 on Build GRCh38
rs191982688 Chr.X: 8791308-8791308 on Build GRCh38
rs763808512 Chr.X: 8791279-8791279 on Build GRCh38
rs750129265 Chr.X: 8791260-8791260 on Build GRCh38
rs1011737814 Chr.X: 8791309-8791309 on Build GRCh38
rs779131302 Chr.X: 8791321-8791321 on Build GRCh38
rs61732571 Chr.X: 8791301-8791301 on Build GRCh38
rs186512412 Chr.X: 8791235-8791235 on Build GRCh38
rs372582389 Chr.X: 8791202-8791202 on Build GRCh38
rs757471549 Chr.X: 8791320-8791320 on Build GRCh38
rs531874464 Chr.X: 8791290-8791290 on Build GRCh38
rs768509456 Chr.X: 8791313-8791313 on Build GRCh38
rs967636529 Chr.X: 8791310-8791310 on Build GRCh38
rs1006915138 Chr.X: 8791179-8791180 on Build GRCh38
rs746133039 Chr.X: 8791325-8791325 on Build GRCh38

View Full Product Details