Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43506733-43506986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETHE1
Entrez Gene ID
23474
Gene Name
ETHE1, persulfide dioxygenase
Gene Aliases
HSCO, YF13H12
UniGene
Hs.7486
Gene Chromosome Location
Chr.19: 43506719-43527256 on Build GRCh38
Transcripts
NM_001320867.1 NM_001320868.1 NM_001320869.1 NM_014297.4 XM_005258687.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
712540 c.732C>T p.N244N Chr.19: 43506883-43506883 on Build GRCh38
3289023 c.736C>T p.R246C Chr.19: 43506879-43506879 on Build GRCh38
4557857 c.744G>A p.G248G Chr.19: 43506871-43506871 on Build GRCh38
997626 c.713-1G>A p.? Chr.19: 43506903-43506903 on Build GRCh38
3535277 c.763T>C p.*255R Chr.19: 43506852-43506852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749242752 Chr.19: 43506814-43506814 on Build GRCh38
rs777652898 Chr.19: 43506881-43506881 on Build GRCh38
rs371701045 Chr.19: 43506946-43506946 on Build GRCh38
rs766509845 Chr.19: 43506870-43506870 on Build GRCh38
rs200273014 Chr.19: 43506904-43506904 on Build GRCh38
rs762443593 Chr.19: 43506855-43506855 on Build GRCh38
rs372169727 Chr.19: 43506820-43506820 on Build GRCh38
rs201943705 Chr.19: 43506935-43506935 on Build GRCh38
rs760882951 Chr.19: 43506843-43506843 on Build GRCh38
rs777509773 Chr.19: 43506802-43506802 on Build GRCh38
rs1055439270 Chr.19: 43506812-43506812 on Build GRCh38
rs765888263 Chr.19: 43506856-43506856 on Build GRCh38
rs201842186 Chr.19: 43506795-43506795 on Build GRCh38
rs901634319 Chr.19: 43506815-43506815 on Build GRCh38
rs549610268 Chr.19: 43506916-43506916 on Build GRCh38
rs930854445 Chr.19: 43506758-43506758 on Build GRCh38
rs939716292 Chr.19: 43506787-43506787 on Build GRCh38
rs777230548 Chr.19: 43506853-43506853 on Build GRCh38
rs763187239 Chr.19: 43506868-43506868 on Build GRCh38
rs770386105 Chr.19: 43506818-43506818 on Build GRCh38
rs753671768 Chr.19: 43506890-43506890 on Build GRCh38
rs751816050 Chr.19: 43506878-43506878 on Build GRCh38
rs895036898 Chr.19: 43506846-43506846 on Build GRCh38
rs747806362 Chr.19: 43506814-43506814 on Build GRCh38
rs780341837 Chr.19: 43506801-43506802 on Build GRCh38
rs892985657 Chr.19: 43506898-43506898 on Build GRCh38
rs878943800 Chr.19: 43506837-43506837 on Build GRCh38
rs1036732356 Chr.19: 43506793-43506793 on Build GRCh38
rs775680830 Chr.19: 43506776-43506776 on Build GRCh38
rs201162412 Chr.19: 43506950-43506950 on Build GRCh38
rs146448778 Chr.19: 43506938-43506938 on Build GRCh38
rs374816177 Chr.19: 43506879-43506879 on Build GRCh38
rs778362220 Chr.19: 43506819-43506819 on Build GRCh38
rs750511915 Chr.19: 43506866-43506866 on Build GRCh38
rs1011887017 Chr.19: 43506912-43506912 on Build GRCh38
rs529990989 Chr.19: 43506800-43506800 on Build GRCh38
rs757163002 Chr.19: 43506909-43506909 on Build GRCh38
rs769079933 Chr.19: 43506844-43506844 on Build GRCh38
rs1020225722 Chr.19: 43506936-43506936 on Build GRCh38
rs531383666 Chr.19: 43506943-43506943 on Build GRCh38
rs779514934 Chr.19: 43506942-43506942 on Build GRCh38
rs775279939 Chr.19: 43506827-43506827 on Build GRCh38

View Full Product Details