Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60764202-60764455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL7
Entrez Gene ID
79993
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
UniGene
Hs.274256
Gene Chromosome Location
Chr.5: 60751789-60844292 on Build GRCh38
Transcripts
NM_001104558.1 NM_001297617.1 NM_001297618.1 NM_024930.2 XM_005248606.4 XM_005248607.3 XM_006714695.3 XM_011543651.2 XM_017009885.1 XM_017009886.1 XM_017009887.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6319606 c.447C>A p.F149L Chr.5: 60764279-60764279 on Build GRCh38
4892583 c.435C>T p.F145F Chr.5: 60764291-60764291 on Build GRCh38
4492367 c.396C>T p.I132I Chr.5: 60764330-60764330 on Build GRCh38
3727743 c.467G>A p.W156* Chr.5: 60764259-60764259 on Build GRCh38
1695812 c.439C>T p.H147Y Chr.5: 60764287-60764287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202133313 Chr.5: 60764261-60764261 on Build GRCh38
rs757660472 Chr.5: 60764232-60764232 on Build GRCh38
rs778418111 Chr.5: 60764344-60764344 on Build GRCh38
rs781370319 Chr.5: 60764242-60764242 on Build GRCh38
rs866345231 Chr.5: 60764263-60764263 on Build GRCh38
rs763099478 Chr.5: 60764313-60764313 on Build GRCh38
rs752879492 Chr.5: 60764309-60764309 on Build GRCh38
rs748991590 Chr.5: 60764245-60764245 on Build GRCh38
rs781629199 Chr.5: 60764330-60764330 on Build GRCh38
rs975977103 Chr.5: 60764290-60764290 on Build GRCh38
rs375929224 Chr.5: 60764286-60764286 on Build GRCh38
rs759820418 Chr.5: 60764299-60764299 on Build GRCh38
rs776243961 Chr.5: 60764297-60764297 on Build GRCh38
rs887878720 Chr.5: 60764328-60764328 on Build GRCh38
rs757872573 Chr.5: 60764370-60764370 on Build GRCh38
rs948841309 Chr.5: 60764346-60764346 on Build GRCh38
rs760504279 Chr.5: 60764274-60764274 on Build GRCh38
rs765312108 Chr.5: 60764307-60764307 on Build GRCh38
rs201510038 Chr.5: 60764339-60764339 on Build GRCh38
rs757103401 Chr.5: 60764333-60764333 on Build GRCh38
rs200379193 Chr.5: 60764317-60764317 on Build GRCh38
rs778488111 Chr.5: 60764255-60764255 on Build GRCh38
rs1044381965 Chr.5: 60764394-60764394 on Build GRCh38
rs773112795 Chr.5: 60764269-60764269 on Build GRCh38
rs752016580 Chr.5: 60764229-60764229 on Build GRCh38
rs200377514 Chr.5: 60764262-60764262 on Build GRCh38
rs764734594 Chr.5: 60764316-60764316 on Build GRCh38
rs777229430 Chr.5: 60764372-60764372 on Build GRCh38
rs768134267 Chr.5: 60764253-60764253 on Build GRCh38
rs757654842 Chr.5: 60764324-60764324 on Build GRCh38
rs928642308 Chr.5: 60764395-60764395 on Build GRCh38

View Full Product Details