Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240594480-240594715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN10
Entrez Gene ID
11132
Gene Name
calpain 10
Gene Aliases
CANP10, NIDDM1
UniGene
Hs.728234
Gene Chromosome Location
Chr.2: 240586716-240599109 on Build GRCh38
Transcripts
NM_023083.3 NM_023085.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4092871 c.969G>C p.E323D Chr.2: 240594681-240594681 on Build GRCh38
6410921 c.997G>T p.E333* Chr.2: 240594709-240594709 on Build GRCh38
6900536 c.861G>A p.A287A Chr.2: 240594573-240594573 on Build GRCh38
1018744 c.896G>A p.G299E Chr.2: 240594608-240594608 on Build GRCh38
6410919 c.997G>T p.E333* Chr.2: 240594709-240594709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146688203 Chr.2: 240594669-240594669 on Build GRCh38
rs761909084 Chr.2: 240594651-240594651 on Build GRCh38
rs932342960 Chr.2: 240594677-240594677 on Build GRCh38
rs760675210 Chr.2: 240594660-240594660 on Build GRCh38
rs137930786 Chr.2: 240594581-240594581 on Build GRCh38
rs777385683 Chr.2: 240594586-240594586 on Build GRCh38
rs17846973 Chr.2: 240594517-240594517 on Build GRCh38
rs1053394682 Chr.2: 240594594-240594594 on Build GRCh38
rs200757356 Chr.2: 240594624-240594624 on Build GRCh38
rs200584759 Chr.2: 240594508-240594508 on Build GRCh38
rs756114074 Chr.2: 240594560-240594560 on Build GRCh38
rs560961041 Chr.2: 240594625-240594625 on Build GRCh38
rs746339773 Chr.2: 240594619-240594619 on Build GRCh38
rs146771809 Chr.2: 240594685-240594685 on Build GRCh38
rs149240512 Chr.2: 240594572-240594572 on Build GRCh38
rs775882611 Chr.2: 240594516-240594516 on Build GRCh38
rs140681762 Chr.2: 240594671-240594671 on Build GRCh38
rs144696781 Chr.2: 240594585-240594585 on Build GRCh38
rs377500913 Chr.2: 240594652-240594652 on Build GRCh38
rs533907687 Chr.2: 240594529-240594529 on Build GRCh38
rs145787041 Chr.2: 240594678-240594678 on Build GRCh38
rs1034936226 Chr.2: 240594598-240594598 on Build GRCh38
rs768811919 Chr.2: 240594645-240594645 on Build GRCh38
rs746599972 Chr.2: 240594682-240594682 on Build GRCh38
rs370216116 Chr.2: 240594545-240594545 on Build GRCh38
rs974232184 Chr.2: 240594673-240594673 on Build GRCh38
rs779969947 Chr.2: 240594562-240594562 on Build GRCh38
rs767436231 Chr.2: 240594558-240594558 on Build GRCh38
rs769036578 Chr.2: 240594692-240594692 on Build GRCh38
rs545243469 Chr.2: 240594643-240594643 on Build GRCh38
rs754673816 Chr.2: 240594573-240594573 on Build GRCh38
rs147221444 Chr.2: 240594583-240594583 on Build GRCh38
rs764369983 Chr.2: 240594539-240594539 on Build GRCh38
rs775831222 Chr.2: 240594630-240594630 on Build GRCh38
rs774455277 Chr.2: 240594647-240594647 on Build GRCh38
rs1040841293 Chr.2: 240594546-240594546 on Build GRCh38
rs778711640 Chr.2: 240594577-240594577 on Build GRCh38
rs17846974 Chr.2: 240594675-240594675 on Build GRCh38
rs201300160 Chr.2: 240594672-240594672 on Build GRCh38
rs771401094 Chr.2: 240594507-240594507 on Build GRCh38
rs749609775 Chr.2: 240594690-240594690 on Build GRCh38
rs371504920 Chr.2: 240594606-240594606 on Build GRCh38
rs893508687 Chr.2: 240594636-240594636 on Build GRCh38
rs757301434 Chr.2: 240594547-240594547 on Build GRCh38
rs780556608 Chr.2: 240594687-240594687 on Build GRCh38
rs766488505 Chr.2: 240594661-240594661 on Build GRCh38
rs750247008 Chr.2: 240594664-240594664 on Build GRCh38

View Full Product Details