Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66313952-66314205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L
Entrez Gene ID
115752
Gene Name
DIS3 like exosome 3'-5' exoribonuclease
Gene Aliases
DIS3L1
UniGene
Hs.446251
Gene Chromosome Location
Chr.15: 66293257-66333898 on Build GRCh38
Transcripts
NM_001143688.2 NM_001323936.1 NM_001323937.1 NM_001323938.1 NM_001323939.1 NM_001323940.1 NM_001323941.1 NM_001323943.1 NM_001323944.1 NM_001323945.1 NM_001323946.1 NM_001323948.1 NM_133375.4 XM_005254146.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
964181 c.789T>G p.L263L Chr.15: 66314092-66314092 on Build GRCh38
964179 c.784C>T p.R262* Chr.15: 66314087-66314087 on Build GRCh38
294312 c.536G>A p.R179Q Chr.15: 66314088-66314088 on Build GRCh38
5691890 c.794G>C p.G265A Chr.15: 66314097-66314097 on Build GRCh38
7429998 c.526G>A p.A176T Chr.15: 66314078-66314078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745375599 Chr.15: 66314024-66314024 on Build GRCh38
rs763174054 Chr.15: 66314087-66314087 on Build GRCh38
rs778429051 Chr.15: 66314020-66314020 on Build GRCh38
rs201218848 Chr.15: 66313994-66313994 on Build GRCh38
rs1051203529 Chr.15: 66314073-66314073 on Build GRCh38
rs778497689 Chr.15: 66314161-66314161 on Build GRCh38
rs770186141 Chr.15: 66314060-66314060 on Build GRCh38
rs745373362 Chr.15: 66314166-66314166 on Build GRCh38
rs756869035 Chr.15: 66314155-66314155 on Build GRCh38
rs774480883 Chr.15: 66314090-66314090 on Build GRCh38
rs369120036 Chr.15: 66314011-66314011 on Build GRCh38
rs777184399 Chr.15: 66314047-66314047 on Build GRCh38
rs868338883 Chr.15: 66314101-66314101 on Build GRCh38
rs990645965 Chr.15: 66314176-66314176 on Build GRCh38
rs767695458 Chr.15: 66314062-66314062 on Build GRCh38
rs139845712 Chr.15: 66314113-66314113 on Build GRCh38
rs748493398 Chr.15: 66314055-66314055 on Build GRCh38
rs372960179 Chr.15: 66314088-66314088 on Build GRCh38
rs780317941 Chr.15: 66314146-66314147 on Build GRCh38
rs373638726 Chr.15: 66314167-66314167 on Build GRCh38
rs924587182 Chr.15: 66313993-66313993 on Build GRCh38
rs769151178 Chr.15: 66314025-66314025 on Build GRCh38
rs767539701 Chr.15: 66314099-66314099 on Build GRCh38
rs1011807399 Chr.15: 66314109-66314109 on Build GRCh38
rs756150223 Chr.15: 66314115-66314115 on Build GRCh38
rs145109090 Chr.15: 66314072-66314072 on Build GRCh38
rs866722540 Chr.15: 66314069-66314069 on Build GRCh38
rs376177769 Chr.15: 66314121-66314121 on Build GRCh38
rs150541699 Chr.15: 66314093-66314093 on Build GRCh38
rs369502331 Chr.15: 66314148-66314148 on Build GRCh38

View Full Product Details