Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40458617-40458852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
UniGene
Hs.22109
Gene Chromosome Location
Chr.15: 40439721-40468242 on Build GRCh38
Transcripts
NM_001301132.1 NM_014952.4 NR_125372.1 XM_011521366.2 XM_011521367.2 XM_011521368.2 XM_011521369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609635 c.227G>A p.R76H Chr.15: 40458691-40458691 on Build GRCh38
961200 c.172C>T p.R58C Chr.15: 40458636-40458636 on Build GRCh38
5722283 c.294G>A p.K98K Chr.15: 40458758-40458758 on Build GRCh38
5772290 c.291C>A p.P97P Chr.15: 40458755-40458755 on Build GRCh38
1517023 c.226C>T p.R76C Chr.15: 40458690-40458690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779561411 Chr.15: 40458767-40458767 on Build GRCh38
rs1015647122 Chr.15: 40458691-40458691 on Build GRCh38
rs768455424 Chr.15: 40458705-40458705 on Build GRCh38
rs753175585 Chr.15: 40458758-40458758 on Build GRCh38
rs763167655 Chr.15: 40458813-40458813 on Build GRCh38
rs1030395511 Chr.15: 40458676-40458676 on Build GRCh38
rs752012097 Chr.15: 40458740-40458740 on Build GRCh38
rs368576045 Chr.15: 40458715-40458715 on Build GRCh38
rs778734004 Chr.15: 40458762-40458762 on Build GRCh38
rs906499797 Chr.15: 40458774-40458774 on Build GRCh38
rs771490327 Chr.15: 40458764-40458764 on Build GRCh38
rs745422765 Chr.15: 40458673-40458673 on Build GRCh38
rs768650125 Chr.15: 40458793-40458793 on Build GRCh38
rs954661448 Chr.15: 40458695-40458695 on Build GRCh38
rs761646925 Chr.15: 40458802-40458802 on Build GRCh38
rs375659522 Chr.15: 40458788-40458788 on Build GRCh38
rs578160651 Chr.15: 40458643-40458643 on Build GRCh38
rs200547065 Chr.15: 40458794-40458794 on Build GRCh38
rs564832968 Chr.15: 40458642-40458642 on Build GRCh38
rs201390167 Chr.15: 40458763-40458763 on Build GRCh38
rs776469369 Chr.15: 40458713-40458713 on Build GRCh38
rs985985022 Chr.15: 40458701-40458701 on Build GRCh38
rs762964387 Chr.15: 40458721-40458721 on Build GRCh38
rs775323463 Chr.15: 40458693-40458693 on Build GRCh38
rs1046422440 Chr.15: 40458770-40458770 on Build GRCh38
rs765066993 Chr.15: 40458826-40458826 on Build GRCh38
rs547045284 Chr.15: 40458804-40458804 on Build GRCh38
rs561606387 Chr.15: 40458769-40458769 on Build GRCh38
rs770762022 Chr.15: 40458647-40458647 on Build GRCh38
rs756512138 Chr.15: 40458760-40458760 on Build GRCh38
rs747097674 Chr.15: 40458757-40458801 on Build GRCh38
rs565157473 Chr.15: 40458707-40458707 on Build GRCh38
rs766440622 Chr.15: 40458732-40458732 on Build GRCh38
rs531314286 Chr.15: 40458752-40458752 on Build GRCh38
rs773337252 Chr.15: 40458807-40458807 on Build GRCh38
rs199747519 Chr.15: 40458714-40458714 on Build GRCh38
rs750621135 Chr.15: 40458717-40458717 on Build GRCh38
rs894054699 Chr.15: 40458800-40458800 on Build GRCh38
rs774137662 Chr.15: 40458654-40458654 on Build GRCh38
rs760597044 Chr.15: 40458702-40458702 on Build GRCh38
rs202044593 Chr.15: 40458743-40458743 on Build GRCh38
rs370119198 Chr.15: 40458681-40458681 on Build GRCh38
rs771566528 Chr.15: 40458685-40458685 on Build GRCh38

View Full Product Details