Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 80504831-80505098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP128
Entrez Gene ID
145508
Gene Name
centrosomal protein 128
Gene Aliases
C14orf145, C14orf61, LEDP/132
UniGene
Hs.162889
Gene Chromosome Location
Chr.14: 80478321-80959501 on Build GRCh38
Transcripts
NM_152446.3 XM_006720056.3 XM_011536490.2 XM_011536491.2 XM_011536492.2 XM_011536493.2 XM_017021042.1 XM_017021043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
317062 c.3073G>T p.G1025C Chr.14: 80505020-80505020 on Build GRCh38
6293462 c.3175G>A p.V1059M Chr.14: 80504918-80504918 on Build GRCh38
4849727 c.3122C>T p.S1041L Chr.14: 80504971-80504971 on Build GRCh38
5964571 c.3175G>T p.V1059L Chr.14: 80504918-80504918 on Build GRCh38
4852287 c.3168T>G p.F1056L Chr.14: 80504925-80504925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762077854 Chr.14: 80504967-80504967 on Build GRCh38
rs757592655 Chr.14: 80504866-80504866 on Build GRCh38
rs753675587 Chr.14: 80505006-80505030 on Build GRCh38
rs530212340 Chr.14: 80505038-80505038 on Build GRCh38
rs150831081 Chr.14: 80504986-80504986 on Build GRCh38
rs774431690 Chr.14: 80505066-80505066 on Build GRCh38
rs777300675 Chr.14: 80504995-80504995 on Build GRCh38
rs763864395 Chr.14: 80504942-80504946 on Build GRCh38
rs74333779 Chr.14: 80505039-80505039 on Build GRCh38
rs770220669 Chr.14: 80504938-80504938 on Build GRCh38
rs373014765 Chr.14: 80504948-80504948 on Build GRCh38
rs773984752 Chr.14: 80504969-80504969 on Build GRCh38
rs774324551 Chr.14: 80504960-80504960 on Build GRCh38
rs377362585 Chr.14: 80504983-80504983 on Build GRCh38
rs200495907 Chr.14: 80504906-80504906 on Build GRCh38
rs752661407 Chr.14: 80504993-80504993 on Build GRCh38
rs769869419 Chr.14: 80505009-80505009 on Build GRCh38
rs188512111 Chr.14: 80505034-80505034 on Build GRCh38
rs759907802 Chr.14: 80504935-80504935 on Build GRCh38
rs945360146 Chr.14: 80505053-80505053 on Build GRCh38
rs748099196 Chr.14: 80504907-80504907 on Build GRCh38
rs940559824 Chr.14: 80504858-80504858 on Build GRCh38
rs974147447 Chr.14: 80504949-80504949 on Build GRCh38
rs777201166 Chr.14: 80504919-80504919 on Build GRCh38
rs202134302 Chr.14: 80504931-80504931 on Build GRCh38
rs952412885 Chr.14: 80504987-80504987 on Build GRCh38
rs746549440 Chr.14: 80505003-80505003 on Build GRCh38
rs764997226 Chr.14: 80504991-80504991 on Build GRCh38
rs746645316 Chr.14: 80504922-80504922 on Build GRCh38
rs764201565 Chr.14: 80504950-80504950 on Build GRCh38
rs758310850 Chr.14: 80504994-80504994 on Build GRCh38
rs753707756 Chr.14: 80504879-80504879 on Build GRCh38
rs61744330 Chr.14: 80504918-80504918 on Build GRCh38
rs955229261 Chr.14: 80504897-80504897 on Build GRCh38
rs776932905 Chr.14: 80504934-80504934 on Build GRCh38
rs372474618 Chr.14: 80505046-80505046 on Build GRCh38
rs753616765 Chr.14: 80504979-80504979 on Build GRCh38
rs563886156 Chr.14: 80505064-80505064 on Build GRCh38
rs3213968 Chr.14: 80504900-80504900 on Build GRCh38
rs779953795 Chr.14: 80504876-80504876 on Build GRCh38
rs770605079 Chr.14: 80504933-80504933 on Build GRCh38
rs541391458 Chr.14: 80504973-80504973 on Build GRCh38
rs539610461 Chr.14: 80505029-80505029 on Build GRCh38
rs776062756 Chr.14: 80504943-80504943 on Build GRCh38
rs767861364 Chr.14: 80504971-80504971 on Build GRCh38
rs761317180 Chr.14: 80505027-80505027 on Build GRCh38
rs780828444 Chr.14: 80505022-80505022 on Build GRCh38

View Full Product Details