Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74799217-74799454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSK
Entrez Gene ID
1445
Gene Name
c-src tyrosine kinase
Gene Aliases
-
UniGene
Hs.77793
Gene Chromosome Location
Chr.15: 74782064-74803198 on Build GRCh38
Transcripts
NM_001127190.1 NM_004383.2 XM_005254165.4 XM_017021925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6304019 c.389G>A p.R130H Chr.15: 74799418-74799418 on Build GRCh38
701189 c.330C>T p.T110T Chr.15: 74799359-74799359 on Build GRCh38
3794402 c.277G>A p.E93K Chr.15: 74799306-74799306 on Build GRCh38
5054487 c.319C>T p.R107W Chr.15: 74799348-74799348 on Build GRCh38
4601996 c.372_384del13 p.K125fs*62 Chr.15: 74799401-74799413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775009760 Chr.15: 74799281-74799281 on Build GRCh38
rs1033335420 Chr.15: 74799418-74799418 on Build GRCh38
rs780001574 Chr.15: 74799325-74799325 on Build GRCh38
rs767255442 Chr.15: 74799318-74799318 on Build GRCh38
rs2229729 Chr.15: 74799323-74799323 on Build GRCh38
rs751418156 Chr.15: 74799310-74799310 on Build GRCh38
rs774934532 Chr.15: 74799407-74799407 on Build GRCh38
rs538976874 Chr.15: 74799252-74799252 on Build GRCh38
rs979623672 Chr.15: 74799398-74799398 on Build GRCh38
rs747486500 Chr.15: 74799395-74799395 on Build GRCh38
rs139298052 Chr.15: 74799404-74799404 on Build GRCh38
rs762515591 Chr.15: 74799427-74799427 on Build GRCh38
rs763802749 Chr.15: 74799307-74799307 on Build GRCh38
rs551800678 Chr.15: 74799386-74799386 on Build GRCh38
rs1007741728 Chr.15: 74799308-74799308 on Build GRCh38
rs367837559 Chr.15: 74799293-74799293 on Build GRCh38
rs773663905 Chr.15: 74799389-74799389 on Build GRCh38
rs1009684691 Chr.15: 74799347-74799347 on Build GRCh38
rs749379756 Chr.15: 74799326-74799326 on Build GRCh38
rs150653589 Chr.15: 74799361-74799361 on Build GRCh38
rs372019231 Chr.15: 74799383-74799383 on Build GRCh38
rs771405153 Chr.15: 74799399-74799399 on Build GRCh38
rs987781536 Chr.15: 74799436-74799436 on Build GRCh38
rs757011709 Chr.15: 74799311-74799311 on Build GRCh38
rs1021581178 Chr.15: 74799368-74799368 on Build GRCh38
rs755212258 Chr.15: 74799329-74799329 on Build GRCh38
rs372190412 Chr.15: 74799380-74799380 on Build GRCh38
rs750380440 Chr.15: 74799319-74799319 on Build GRCh38
rs761206148 Chr.15: 74799239-74799239 on Build GRCh38

View Full Product Details