Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96203544-96203817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT9
Entrez Gene ID
10690
Gene Name
fucosyltransferase 9
Gene Aliases
Fuc-TIX
UniGene
Hs.49117
Gene Chromosome Location
Chr.6: 96015969-96215612 on Build GRCh38
Transcripts
NM_006581.3 XM_011535383.2 XM_011535385.2 XM_017010188.1 XM_017010190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6175029 c.460C>A p.L154M Chr.6: 96203615-96203615 on Build GRCh38
3922268 c.529C>T p.P177S Chr.6: 96203684-96203684 on Build GRCh38
4852365 c.645C>G p.I215M Chr.6: 96203800-96203800 on Build GRCh38
3876228 c.466C>G p.L156V Chr.6: 96203621-96203621 on Build GRCh38
3630653 c.640G>A p.E214K Chr.6: 96203795-96203795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527639500 Chr.6: 96203633-96203633 on Build GRCh38
rs766706271 Chr.6: 96203746-96203746 on Build GRCh38
rs775402689 Chr.6: 96203581-96203581 on Build GRCh38
rs765735833 Chr.6: 96203630-96203630 on Build GRCh38
rs760109451 Chr.6: 96203614-96203614 on Build GRCh38
rs745684001 Chr.6: 96203672-96203672 on Build GRCh38
rs777335027 Chr.6: 96203617-96203617 on Build GRCh38
rs770446854 Chr.6: 96203720-96203720 on Build GRCh38
rs779588196 Chr.6: 96203689-96203689 on Build GRCh38
rs763326668 Chr.6: 96203634-96203634 on Build GRCh38
rs375082399 Chr.6: 96203686-96203686 on Build GRCh38
rs752800996 Chr.6: 96203790-96203790 on Build GRCh38
rs936441917 Chr.6: 96203776-96203776 on Build GRCh38
rs767637286 Chr.6: 96203778-96203778 on Build GRCh38
rs766540242 Chr.6: 96203640-96203640 on Build GRCh38
rs371085883 Chr.6: 96203648-96203648 on Build GRCh38
rs549928588 Chr.6: 96203725-96203725 on Build GRCh38
rs1042257798 Chr.6: 96203740-96203740 on Build GRCh38
rs541189278 Chr.6: 96203644-96203644 on Build GRCh38
rs778777463 Chr.6: 96203671-96203671 on Build GRCh38
rs367998184 Chr.6: 96203732-96203732 on Build GRCh38
rs201945905 Chr.6: 96203692-96203692 on Build GRCh38
rs753835629 Chr.6: 96203665-96203665 on Build GRCh38
rs989822927 Chr.6: 96203764-96203764 on Build GRCh38
rs151138332 Chr.6: 96203587-96203587 on Build GRCh38
rs142240115 Chr.6: 96203584-96203584 on Build GRCh38
rs199802287 Chr.6: 96203748-96203748 on Build GRCh38
rs368301077 Chr.6: 96203608-96203608 on Build GRCh38
rs771647857 Chr.6: 96203592-96203592 on Build GRCh38
rs768225277 Chr.6: 96203615-96203615 on Build GRCh38
rs759746109 Chr.6: 96203762-96203762 on Build GRCh38
rs746661735 Chr.6: 96203690-96203690 on Build GRCh38
rs867214750 Chr.6: 96203685-96203685 on Build GRCh38
rs762458149 Chr.6: 96203717-96203717 on Build GRCh38
rs757168999 Chr.6: 96203670-96203670 on Build GRCh38
rs763230210 Chr.6: 96203734-96203734 on Build GRCh38
rs775171320 Chr.6: 96203596-96203596 on Build GRCh38
rs781231611 Chr.6: 96203654-96203654 on Build GRCh38

View Full Product Details