Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 82733421-82733646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM181B
Entrez Gene ID
220382
Gene Name
family with sequence similarity 181 member B
Gene Aliases
-
UniGene
Hs.448218
Gene Chromosome Location
Chr.11: 82732004-82733864 on Build GRCh38
Transcripts
NM_175885.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6400568 c.139G>T p.A47S Chr.11: 82733591-82733591 on Build GRCh38
1746606 c.289C>T p.L97L Chr.11: 82733441-82733441 on Build GRCh38
7332048 c.87C>T p.F29F Chr.11: 82733643-82733643 on Build GRCh38
5051168 c.153G>A p.L51L Chr.11: 82733577-82733577 on Build GRCh38
4037054 c.108C>T p.C36C Chr.11: 82733622-82733622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763711151 Chr.11: 82733581-82733581 on Build GRCh38
rs779483783 Chr.11: 82733514-82733514 on Build GRCh38
rs759417811 Chr.11: 82733556-82733556 on Build GRCh38
rs370272011 Chr.11: 82733561-82733561 on Build GRCh38
rs773906493 Chr.11: 82733616-82733616 on Build GRCh38
rs772224751 Chr.11: 82733532-82733532 on Build GRCh38
rs763476470 Chr.11: 82733477-82733477 on Build GRCh38
rs972338601 Chr.11: 82733627-82733627 on Build GRCh38
rs747073876 Chr.11: 82733601-82733601 on Build GRCh38
rs747282401 Chr.11: 82733550-82733550 on Build GRCh38
rs963848892 Chr.11: 82733516-82733516 on Build GRCh38
rs781569097 Chr.11: 82733604-82733604 on Build GRCh38
rs377416489 Chr.11: 82733442-82733442 on Build GRCh38
rs776543693 Chr.11: 82733461-82733461 on Build GRCh38
rs368475342 Chr.11: 82733602-82733602 on Build GRCh38
rs777948254 Chr.11: 82733598-82733598 on Build GRCh38
rs759615699 Chr.11: 82733463-82733463 on Build GRCh38
rs767140869 Chr.11: 82733589-82733589 on Build GRCh38
rs757934980 Chr.11: 82733473-82733473 on Build GRCh38
rs371315457 Chr.11: 82733609-82733609 on Build GRCh38
rs1010580111 Chr.11: 82733515-82733515 on Build GRCh38
rs745663474 Chr.11: 82733542-82733553 on Build GRCh38
rs893460753 Chr.11: 82733492-82733492 on Build GRCh38
rs780433938 Chr.11: 82733499-82733499 on Build GRCh38
rs768471286 Chr.11: 82733610-82733610 on Build GRCh38
rs752520156 Chr.11: 82733471-82733471 on Build GRCh38
rs758807400 Chr.11: 82733597-82733597 on Build GRCh38
rs755532194 Chr.11: 82733510-82733510 on Build GRCh38
rs756966935 Chr.11: 82733586-82733586 on Build GRCh38
rs766298977 Chr.11: 82733460-82733460 on Build GRCh38
rs762755229 Chr.11: 82733564-82733564 on Build GRCh38
rs148661098 Chr.11: 82733560-82733560 on Build GRCh38
rs773115800 Chr.11: 82733543-82733543 on Build GRCh38
rs753268924 Chr.11: 82733595-82733595 on Build GRCh38
rs144169331 Chr.11: 82733490-82733490 on Build GRCh38
rs765253866 Chr.11: 82733465-82733465 on Build GRCh38
rs761322070 Chr.11: 82733621-82733621 on Build GRCh38
rs760768505 Chr.11: 82733454-82733454 on Build GRCh38
rs750016052 Chr.11: 82733590-82733590 on Build GRCh38
rs751249446 Chr.11: 82733481-82733481 on Build GRCh38
rs771275042 Chr.11: 82733551-82733551 on Build GRCh38
rs376354005 Chr.11: 82733592-82733592 on Build GRCh38
rs755336409 Chr.11: 82733591-82733591 on Build GRCh38
rs144429777 Chr.11: 82733626-82733626 on Build GRCh38
rs751005808 Chr.11: 82733583-82733583 on Build GRCh38
rs1029439982 Chr.11: 82733571-82733571 on Build GRCh38
rs746050994 Chr.11: 82733607-82733607 on Build GRCh38
rs776627689 Chr.11: 82733552-82733552 on Build GRCh38

View Full Product Details