Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77196745-77197018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC146
Entrez Gene ID
57639
Gene Name
coiled-coil domain containing 146
Gene Aliases
-
UniGene
Hs.113940
Gene Chromosome Location
Chr.7: 77122617-77295204 on Build GRCh38
Transcripts
NM_020879.2 NM_006682.2

Gene Symbol
FGL2
Entrez Gene ID
10875
Gene Name
fibrinogen like 2
Gene Aliases
T49, pT49
UniGene
Hs.520989
Gene Chromosome Location
Chr.7: 77193371-77199833 on Build GRCh38
Transcripts
NM_020879.2 NM_006682.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4684642 c.769C>T p.R257C Chr.7: 77196830-77196830 on Build GRCh38
4632441 c.851T>C p.L284P Chr.7: 77196748-77196748 on Build GRCh38
3923977 c.673G>A p.E225K Chr.7: 77196926-77196926 on Build GRCh38
3640690 c.720T>G p.V240V Chr.7: 77196879-77196879 on Build GRCh38
6484649 c.818C>G p.A273G Chr.7: 77196781-77196781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548765525 Chr.7: 77196858-77196858 on Build GRCh38
rs780583784 Chr.7: 77196939-77196939 on Build GRCh38
rs753328837 Chr.7: 77196798-77196798 on Build GRCh38
rs1016961575 Chr.7: 77196817-77196817 on Build GRCh38
rs147689299 Chr.7: 77196933-77196933 on Build GRCh38
rs770119287 Chr.7: 77196855-77196855 on Build GRCh38
rs760659595 Chr.7: 77196903-77196903 on Build GRCh38
rs987971335 Chr.7: 77196980-77196980 on Build GRCh38
rs555120974 Chr.7: 77196987-77196987 on Build GRCh38
rs185392374 Chr.7: 77196962-77196962 on Build GRCh38
rs777734007 Chr.7: 77196834-77196834 on Build GRCh38
rs751564138 Chr.7: 77196934-77196935 on Build GRCh38
rs749496298 Chr.7: 77196971-77196971 on Build GRCh38
rs758025210 Chr.7: 77196937-77196937 on Build GRCh38
rs374306688 Chr.7: 77196829-77196829 on Build GRCh38
rs1013266693 Chr.7: 77196895-77196895 on Build GRCh38
rs147079564 Chr.7: 77196941-77196941 on Build GRCh38
rs775308310 Chr.7: 77196975-77196975 on Build GRCh38
rs267601572 Chr.7: 77196981-77196981 on Build GRCh38
rs566096585 Chr.7: 77196948-77196948 on Build GRCh38
rs775217486 Chr.7: 77196882-77196882 on Build GRCh38
rs371989577 Chr.7: 77196922-77196922 on Build GRCh38
rs368142215 Chr.7: 77196838-77196838 on Build GRCh38
rs769924412 Chr.7: 77196947-77196947 on Build GRCh38
rs748206225 Chr.7: 77196843-77196843 on Build GRCh38
rs756977540 Chr.7: 77196800-77196800 on Build GRCh38
rs994124675 Chr.7: 77196870-77196870 on Build GRCh38
rs777871688 Chr.7: 77196970-77196970 on Build GRCh38
rs778428328 Chr.7: 77196822-77196822 on Build GRCh38
rs138329092 Chr.7: 77196958-77196958 on Build GRCh38
rs201777914 Chr.7: 77196825-77196825 on Build GRCh38
rs771120137 Chr.7: 77196972-77196972 on Build GRCh38
rs1005562886 Chr.7: 77196783-77196783 on Build GRCh38
rs773321059 Chr.7: 77196856-77196856 on Build GRCh38
rs780495796 Chr.7: 77196830-77196830 on Build GRCh38
rs781470633 Chr.7: 77196839-77196839 on Build GRCh38
rs748315331 Chr.7: 77196949-77196949 on Build GRCh38
rs764744247 Chr.7: 77196930-77196930 on Build GRCh38
rs960459619 Chr.7: 77196869-77196869 on Build GRCh38
rs761218758 Chr.7: 77196928-77196928 on Build GRCh38

View Full Product Details