Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40554630-40554899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR7
Entrez Gene ID
1236
Gene Name
C-C motif chemokine receptor 7
Gene Aliases
BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1
UniGene
Hs.370036
Gene Chromosome Location
Chr.17: 40553769-40565484 on Build GRCh38
Transcripts
NM_001301714.1 NM_001301716.1 NM_001301717.1 NM_001301718.1 NM_001838.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670291 c.1085G>A p.R362Q Chr.17: 40554794-40554794 on Build GRCh38
5706377 c.1023delC p.F341fs*26 Chr.17: 40554856-40554856 on Build GRCh38
5706376 c.1045C>T p.Q349* Chr.17: 40554834-40554834 on Build GRCh38
3517062 c.1056C>A p.L352L Chr.17: 40554823-40554823 on Build GRCh38
4670292 c.1084C>T p.R362W Chr.17: 40554795-40554795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778527213 Chr.17: 40554808-40554808 on Build GRCh38
rs371882270 Chr.17: 40554740-40554740 on Build GRCh38
rs142089801 Chr.17: 40554868-40554868 on Build GRCh38
rs200794532 Chr.17: 40554821-40554821 on Build GRCh38
rs1010575135 Chr.17: 40554659-40554659 on Build GRCh38
rs767647420 Chr.17: 40554787-40554787 on Build GRCh38
rs150068770 Chr.17: 40554795-40554795 on Build GRCh38
rs991139043 Chr.17: 40554781-40554781 on Build GRCh38
rs976794673 Chr.17: 40554850-40554850 on Build GRCh38
rs769732065 Chr.17: 40554849-40554849 on Build GRCh38
rs771649965 Chr.17: 40554708-40554708 on Build GRCh38
rs146694134 Chr.17: 40554879-40554879 on Build GRCh38
rs368660693 Chr.17: 40554725-40554725 on Build GRCh38
rs774839512 Chr.17: 40554731-40554731 on Build GRCh38
rs778685035 Chr.17: 40554701-40554701 on Build GRCh38
rs915570645 Chr.17: 40554783-40554783 on Build GRCh38
rs554704609 Chr.17: 40554680-40554680 on Build GRCh38
rs148614545 Chr.17: 40554863-40554863 on Build GRCh38
rs774431889 Chr.17: 40554852-40554852 on Build GRCh38
rs761077309 Chr.17: 40554870-40554870 on Build GRCh38
rs377519447 Chr.17: 40554811-40554811 on Build GRCh38
rs372376814 Chr.17: 40554739-40554739 on Build GRCh38
rs747787289 Chr.17: 40554706-40554706 on Build GRCh38
rs1016439158 Chr.17: 40554690-40554690 on Build GRCh38
rs962481343 Chr.17: 40554716-40554716 on Build GRCh38
rs761858491 Chr.17: 40554853-40554853 on Build GRCh38
rs1003291228 Chr.17: 40554658-40554658 on Build GRCh38
rs762029652 Chr.17: 40554769-40554769 on Build GRCh38
rs770764441 Chr.17: 40554832-40554832 on Build GRCh38
rs1006873994 Chr.17: 40554684-40554684 on Build GRCh38
rs747001394 Chr.17: 40554652-40554652 on Build GRCh38
rs746791164 Chr.17: 40554735-40554735 on Build GRCh38
rs901953581 Chr.17: 40554805-40554805 on Build GRCh38
rs143854738 Chr.17: 40554803-40554803 on Build GRCh38
rs746812817 Chr.17: 40554822-40554822 on Build GRCh38
rs761014378 Chr.17: 40554794-40554794 on Build GRCh38
rs760095834 Chr.17: 40554790-40554790 on Build GRCh38
rs372738708 Chr.17: 40554766-40554766 on Build GRCh38
rs745769227 Chr.17: 40554844-40554844 on Build GRCh38
rs924489237 Chr.17: 40554796-40554796 on Build GRCh38
rs895973358 Chr.17: 40554695-40554695 on Build GRCh38
rs534538889 Chr.17: 40554817-40554817 on Build GRCh38
rs755563290 Chr.17: 40554804-40554804 on Build GRCh38
rs200037144 Chr.17: 40554818-40554818 on Build GRCh38
rs374797000 Chr.17: 40554791-40554791 on Build GRCh38
rs758006926 Chr.17: 40554816-40554816 on Build GRCh38
rs754285590 Chr.17: 40554875-40554875 on Build GRCh38
rs947333410 Chr.17: 40554792-40554792 on Build GRCh38
rs140172763 Chr.17: 40554754-40554754 on Build GRCh38
rs766799198 Chr.17: 40554874-40554874 on Build GRCh38
rs181966581 Chr.17: 40554697-40554697 on Build GRCh38
rs772407156 Chr.17: 40554797-40554797 on Build GRCh38
rs763002099 Chr.17: 40554750-40554750 on Build GRCh38
rs780990133 Chr.17: 40554836-40554836 on Build GRCh38

View Full Product Details