Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34397424-34397697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf24
Entrez Gene ID
84688
Gene Name
chromosome 9 open reading frame 24
Gene Aliases
CBE1, NYD-SP22, SMRP1, bA573M23.4
UniGene
Hs.50334
Gene Chromosome Location
Chr.9: 34379019-34400833 on Build GRCh38
Transcripts
NM_032596.3 XM_005251616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7137373 c.8G>C p.W3S Chr.9: 34397522-34397522 on Build GRCh38
3657019 c.108C>T p.A36A Chr.9: 34397526-34397526 on Build GRCh38
1199090 c.17G>A p.R6H Chr.9: 34397617-34397617 on Build GRCh38
3926722 c.100C>T p.L34L Chr.9: 34397534-34397534 on Build GRCh38
5794356 c.22A>C p.T8P Chr.9: 34397612-34397612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776095615 Chr.9: 34397485-34397485 on Build GRCh38
rs547588201 Chr.9: 34397517-34397517 on Build GRCh38
rs774635037 Chr.9: 34397634-34397635 on Build GRCh38
rs780015404 Chr.9: 34397448-34397448 on Build GRCh38
rs527976842 Chr.9: 34397486-34397486 on Build GRCh38
rs780254101 Chr.9: 34397473-34397473 on Build GRCh38
rs764530738 Chr.9: 34397494-34397494 on Build GRCh38
rs756236763 Chr.9: 34397503-34397503 on Build GRCh38
rs927129209 Chr.9: 34397589-34397589 on Build GRCh38
rs750575497 Chr.9: 34397502-34397502 on Build GRCh38
rs760210728 Chr.9: 34397483-34397483 on Build GRCh38
rs749610698 Chr.9: 34397656-34397656 on Build GRCh38
rs772751956 Chr.9: 34397474-34397474 on Build GRCh38
rs374001098 Chr.9: 34397613-34397613 on Build GRCh38
rs754861400 Chr.9: 34397522-34397522 on Build GRCh38
rs376270101 Chr.9: 34397557-34397557 on Build GRCh38
rs757617269 Chr.9: 34397499-34397499 on Build GRCh38
rs767941024 Chr.9: 34397628-34397628 on Build GRCh38
rs775944639 Chr.9: 34397577-34397577 on Build GRCh38
rs768506503 Chr.9: 34397459-34397459 on Build GRCh38
rs747908462 Chr.9: 34397537-34397537 on Build GRCh38
rs766541000 Chr.9: 34397651-34397651 on Build GRCh38
rs758221899 Chr.9: 34397673-34397673 on Build GRCh38
rs11790577 Chr.9: 34397547-34397547 on Build GRCh38
rs752584359 Chr.9: 34397672-34397672 on Build GRCh38
rs777426211 Chr.9: 34397553-34397553 on Build GRCh38
rs753862813 Chr.9: 34397653-34397653 on Build GRCh38
rs145985963 Chr.9: 34397618-34397618 on Build GRCh38
rs769024447 Chr.9: 34397553-34397554 on Build GRCh38
rs184877012 Chr.9: 34397524-34397524 on Build GRCh38
rs754984120 Chr.9: 34397654-34397654 on Build GRCh38
rs988905947 Chr.9: 34397550-34397550 on Build GRCh38
rs749714414 Chr.9: 34397516-34397517 on Build GRCh38
rs757710374 Chr.9: 34397661-34397661 on Build GRCh38
rs746601224 Chr.9: 34397555-34397555 on Build GRCh38
rs778748576 Chr.9: 34397530-34397530 on Build GRCh38
rs753831851 Chr.9: 34397519-34397519 on Build GRCh38
rs747859243 Chr.9: 34397469-34397469 on Build GRCh38
rs774077596 Chr.9: 34397461-34397461 on Build GRCh38
rs762241532 Chr.9: 34397626-34397626 on Build GRCh38
rs751957678 Chr.9: 34397498-34397498 on Build GRCh38
rs765759062 Chr.9: 34397484-34397484 on Build GRCh38
rs769279212 Chr.9: 34397615-34397615 on Build GRCh38
rs549977241 Chr.9: 34397637-34397637 on Build GRCh38
rs959832269 Chr.9: 34397603-34397603 on Build GRCh38
rs141607377 Chr.9: 34397581-34397581 on Build GRCh38
rs751113995 Chr.9: 34397470-34397470 on Build GRCh38
rs767814035 Chr.9: 34397500-34397500 on Build GRCh38
rs750666209 Chr.9: 34397636-34397636 on Build GRCh38
rs138544446 Chr.9: 34397572-34397572 on Build GRCh38
rs749143134 Chr.9: 34397457-34397457 on Build GRCh38

View Full Product Details