Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43809700-43809933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG5
Entrez Gene ID
64240
Gene Name
ATP binding cassette subfamily G member 5
Gene Aliases
STSL
UniGene
Hs.132992
Gene Chromosome Location
Chr.2: 43806154-43838840 on Build GRCh38
Transcripts
XM_006712073.3 NM_001193464.1 NM_016008.3 XM_005264364.4 XM_005264365.4

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
XM_006712073.3 NM_001193464.1 NM_016008.3 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316792 c.1032G>A p.W344* Chr.2: 43809743-43809743 on Build GRCh38
297110 c.1010A>C p.K337T Chr.2: 43809721-43809721 on Build GRCh38
1306813 c.1048G>C p.D350H Chr.2: 43809759-43809759 on Build GRCh38
3184957 c.1041C>T p.I347I Chr.2: 43809752-43809752 on Build GRCh38
5465567 c.1013G>T p.R338I Chr.2: 43809724-43809724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775767441 Chr.2: 43809786-43809786 on Build GRCh38
rs17031643 Chr.2: 43809908-43809908 on Build GRCh38
rs8302 Chr.2: 43809774-43809774 on Build GRCh38
rs41281457 Chr.2: 43809753-43809753 on Build GRCh38
rs774738890 Chr.2: 43809809-43809809 on Build GRCh38
rs780620194 Chr.2: 43809812-43809812 on Build GRCh38
rs11556156 Chr.2: 43809906-43809906 on Build GRCh38
rs762046678 Chr.2: 43809813-43809813 on Build GRCh38
rs11556154 Chr.2: 43809884-43809884 on Build GRCh38
rs747785913 Chr.2: 43809751-43809751 on Build GRCh38
rs527385102 Chr.2: 43809836-43809836 on Build GRCh38
rs374650741 Chr.2: 43809752-43809752 on Build GRCh38
rs1028226581 Chr.2: 43809784-43809784 on Build GRCh38
rs954845387 Chr.2: 43809838-43809838 on Build GRCh38
rs200850212 Chr.2: 43809763-43809763 on Build GRCh38
rs372677433 Chr.2: 43809778-43809778 on Build GRCh38
rs772980227 Chr.2: 43809824-43809824 on Build GRCh38
rs983968905 Chr.2: 43809814-43809814 on Build GRCh38
rs561404332 Chr.2: 43809807-43809807 on Build GRCh38
rs11556155 Chr.2: 43809799-43809799 on Build GRCh38
rs10186552 Chr.2: 43809783-43809783 on Build GRCh38
rs953920981 Chr.2: 43809793-43809793 on Build GRCh38
rs763239509 Chr.2: 43809791-43809791 on Build GRCh38

View Full Product Details