Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130376035-130376304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A5
Entrez Gene ID
256076
Gene Name
collagen type VI alpha 5
Gene Aliases
COL29A1, VWA4
UniGene
Hs.205403
Gene Chromosome Location
Chr.3: 130345516-130484846 on Build GRCh38
Transcripts
NM_001278298.1 NM_153264.6 NR_022012.2 XM_011512621.2 XM_011512622.2 XM_011512623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1038162 c.115G>A p.D39N Chr.3: 130376284-130376284 on Build GRCh38
1038161 c.93C>T p.V31V Chr.3: 130376262-130376262 on Build GRCh38
4427692 c.72A>T p.P24P Chr.3: 130376241-130376241 on Build GRCh38
1418874 c.133T>C p.S45P Chr.3: 130376302-130376302 on Build GRCh38
5934729 c.77C>T p.P26L Chr.3: 130376246-130376246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051521498 Chr.3: 130376174-130376174 on Build GRCh38
rs993923777 Chr.3: 130376240-130376240 on Build GRCh38
rs758018910 Chr.3: 130376247-130376247 on Build GRCh38
rs754888354 Chr.3: 130376234-130376234 on Build GRCh38
rs760536528 Chr.3: 130376222-130376222 on Build GRCh38
rs997762709 Chr.3: 130376185-130376185 on Build GRCh38
rs9835065 Chr.3: 130376188-130376188 on Build GRCh38
rs148536866 Chr.3: 130376205-130376205 on Build GRCh38
rs186855876 Chr.3: 130376156-130376156 on Build GRCh38
rs369620303 Chr.3: 130376283-130376283 on Build GRCh38
rs749769145 Chr.3: 130376268-130376268 on Build GRCh38
rs769061297 Chr.3: 130376273-130376273 on Build GRCh38
rs375962998 Chr.3: 130376262-130376262 on Build GRCh38
rs749593467 Chr.3: 130376199-130376199 on Build GRCh38
rs142846354 Chr.3: 130376236-130376236 on Build GRCh38
rs151042455 Chr.3: 130376241-130376241 on Build GRCh38
rs193157516 Chr.3: 130376178-130376178 on Build GRCh38
rs574911572 Chr.3: 130376062-130376062 on Build GRCh38
rs1036494070 Chr.3: 130376150-130376150 on Build GRCh38
rs777176863 Chr.3: 130376250-130376250 on Build GRCh38
rs76578069 Chr.3: 130376153-130376153 on Build GRCh38
rs75943025 Chr.3: 130376095-130376095 on Build GRCh38
rs773305489 Chr.3: 130376219-130376219 on Build GRCh38
rs887878645 Chr.3: 130376089-130376089 on Build GRCh38
rs541973078 Chr.3: 130376074-130376074 on Build GRCh38
rs766362600 Chr.3: 130376227-130376227 on Build GRCh38
rs928641215 Chr.3: 130376243-130376243 on Build GRCh38
rs1020164563 Chr.3: 130376233-130376233 on Build GRCh38
rs774480503 Chr.3: 130376209-130376209 on Build GRCh38
rs936118185 Chr.3: 130376077-130376077 on Build GRCh38
rs770243214 Chr.3: 130376187-130376187 on Build GRCh38
rs892039253 Chr.3: 130376101-130376101 on Build GRCh38
rs1006570970 Chr.3: 130376192-130376192 on Build GRCh38
rs746459908 Chr.3: 130376258-130376258 on Build GRCh38
rs531010650 Chr.3: 130376098-130376098 on Build GRCh38
rs762076745 Chr.3: 130376210-130376210 on Build GRCh38
rs780591576 Chr.3: 130376263-130376263 on Build GRCh38

View Full Product Details