Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72822447-72822690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG16L2
Entrez Gene ID
89849
Gene Name
autophagy related 16 like 2
Gene Aliases
ATG16B, WDR80
UniGene
Hs.653186
Gene Chromosome Location
Chr.11: 72814406-72843740 on Build GRCh38
Transcripts
NM_001318766.1 NM_033388.1 XM_005274376.4 XM_006718732.2 XM_006718733.3 XM_006718734.2 XM_011545332.1 XM_011545333.1 XM_011545334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356807 c.701G>T p.S234I Chr.11: 72822534-72822534 on Build GRCh38
5349728 c.675G>A p.L225L Chr.11: 72822508-72822508 on Build GRCh38
6294335 c.659G>T p.R220L Chr.11: 72822492-72822492 on Build GRCh38
4387739 c.687C>A p.A229A Chr.11: 72822520-72822520 on Build GRCh38
6345296 c.682G>T p.A228S Chr.11: 72822515-72822515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775135083 Chr.11: 72822529-72822529 on Build GRCh38
rs774998623 Chr.11: 72822508-72822510 on Build GRCh38
rs868708826 Chr.11: 72822603-72822603 on Build GRCh38
rs746527281 Chr.11: 72822594-72822594 on Build GRCh38
rs772746188 Chr.11: 72822485-72822485 on Build GRCh38
rs771688111 Chr.11: 72822484-72822484 on Build GRCh38
rs767819746 Chr.11: 72822556-72822556 on Build GRCh38
rs374108476 Chr.11: 72822470-72822470 on Build GRCh38
rs759253273 Chr.11: 72822512-72822512 on Build GRCh38
rs1018657477 Chr.11: 72822639-72822639 on Build GRCh38
rs750550696 Chr.11: 72822562-72822562 on Build GRCh38
rs777937554 Chr.11: 72822579-72822580 on Build GRCh38
rs551946729 Chr.11: 72822634-72822634 on Build GRCh38
rs148743255 Chr.11: 72822524-72822524 on Build GRCh38
rs185295626 Chr.11: 72822471-72822471 on Build GRCh38
rs374500366 Chr.11: 72822581-72822587 on Build GRCh38
rs200456427 Chr.11: 72822489-72822489 on Build GRCh38
rs934635019 Chr.11: 72822638-72822638 on Build GRCh38
rs973834938 Chr.11: 72822563-72822563 on Build GRCh38
rs4944806 Chr.11: 72822666-72822666 on Build GRCh38
rs376832672 Chr.11: 72822573-72822573 on Build GRCh38
rs889061472 Chr.11: 72822623-72822623 on Build GRCh38
rs746459407 Chr.11: 72822573-72822574 on Build GRCh38
rs758070793 Chr.11: 72822582-72822582 on Build GRCh38
rs777366428 Chr.11: 72822590-72822590 on Build GRCh38
rs533399984 Chr.11: 72822608-72822608 on Build GRCh38
rs1020684549 Chr.11: 72822670-72822670 on Build GRCh38
rs754309110 Chr.11: 72822568-72822568 on Build GRCh38
rs776573775 Chr.11: 72822509-72822509 on Build GRCh38
rs746480218 Chr.11: 72822490-72822490 on Build GRCh38
rs139902397 Chr.11: 72822580-72822586 on Build GRCh38
rs11235604 Chr.11: 72822491-72822491 on Build GRCh38
rs1040564830 Chr.11: 72822629-72822629 on Build GRCh38
rs753040414 Chr.11: 72822580-72822580 on Build GRCh38
rs747889378 Chr.11: 72822474-72822474 on Build GRCh38
rs370540910 Chr.11: 72822567-72822567 on Build GRCh38
rs769697470 Chr.11: 72822513-72822513 on Build GRCh38
rs141490660 Chr.11: 72822532-72822532 on Build GRCh38
rs779378328 Chr.11: 72822577-72822577 on Build GRCh38
rs200280731 Chr.11: 72822579-72822584 on Build GRCh38
rs965700025 Chr.11: 72822645-72822645 on Build GRCh38
rs771298486 Chr.11: 72822471-72822473 on Build GRCh38

View Full Product Details