Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95445688-95445945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749053 c.1622G>A p.S541N Chr.14: 95445919-95445919 on Build GRCh38
3499517 c.1608C>T p.L536L Chr.14: 95445933-95445933 on Build GRCh38
1749052 c.1622G>A p.S541N Chr.14: 95445919-95445919 on Build GRCh38
3499518 c.1608C>T p.L536L Chr.14: 95445933-95445933 on Build GRCh38
7112218 c.1589_1604del16 p.D530fs*132 Chr.14: 95445937-95445952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201457491 Chr.14: 95445886-95445886 on Build GRCh38
rs529968830 Chr.14: 95445828-95445828 on Build GRCh38
rs903882087 Chr.14: 95445746-95445746 on Build GRCh38
rs543989185 Chr.14: 95445763-95445763 on Build GRCh38
rs8013258 Chr.14: 95445835-95445835 on Build GRCh38
rs888006766 Chr.14: 95445809-95445809 on Build GRCh38
rs769835640 Chr.14: 95445884-95445884 on Build GRCh38
rs753472632 Chr.14: 95445905-95445905 on Build GRCh38
rs201334821 Chr.14: 95445885-95445885 on Build GRCh38
rs377117691 Chr.14: 95445902-95445902 on Build GRCh38
rs17092280 Chr.14: 95445719-95445719 on Build GRCh38
rs781323001 Chr.14: 95445872-95445872 on Build GRCh38
rs140803073 Chr.14: 95445858-95445858 on Build GRCh38
rs556818166 Chr.14: 95445921-95445921 on Build GRCh38
rs73339172 Chr.14: 95445867-95445867 on Build GRCh38
rs151088038 Chr.14: 95445808-95445808 on Build GRCh38
rs114103317 Chr.14: 95445891-95445891 on Build GRCh38
rs764896066 Chr.14: 95445916-95445916 on Build GRCh38
rs894593949 Chr.14: 95445715-95445715 on Build GRCh38
rs746076179 Chr.14: 95445882-95445882 on Build GRCh38
rs769569605 Chr.14: 95445894-95445894 on Build GRCh38
rs1045709646 Chr.14: 95445723-95445723 on Build GRCh38
rs759221646 Chr.14: 95445908-95445908 on Build GRCh38
rs993722962 Chr.14: 95445790-95445790 on Build GRCh38
rs1023800883 Chr.14: 95445843-95445843 on Build GRCh38
rs1018180203 Chr.14: 95445837-95445837 on Build GRCh38
rs975807556 Chr.14: 95445866-95445866 on Build GRCh38

View Full Product Details