Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8605862-8606091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPZ
Entrez Gene ID
8532
Gene Name
carboxypeptidase Z
Gene Aliases
-
UniGene
Hs.61995
Gene Chromosome Location
Chr.4: 8592660-8619761 on Build GRCh38
Transcripts
NM_001014447.2 NM_001014448.2 NM_003652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
149639 c.758C>T p.A253V Chr.4: 8606037-8606037 on Build GRCh38
4819956 c.751G>A p.E251K Chr.4: 8606030-8606030 on Build GRCh38
6345899 c.771G>T p.M257I Chr.4: 8606050-8606050 on Build GRCh38
7259737 c.811C>A p.L271I Chr.4: 8606090-8606090 on Build GRCh38
3993828 c.795G>C p.L265L Chr.4: 8606074-8606074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746298338 Chr.4: 8605958-8605958 on Build GRCh38
rs202043630 Chr.4: 8606022-8606022 on Build GRCh38
rs994823305 Chr.4: 8606009-8606009 on Build GRCh38
rs920475771 Chr.4: 8605987-8605987 on Build GRCh38
rs772142122 Chr.4: 8606068-8606068 on Build GRCh38
rs776226356 Chr.4: 8605972-8605975 on Build GRCh38
rs927819827 Chr.4: 8605900-8605900 on Build GRCh38
rs111373464 Chr.4: 8605922-8605922 on Build GRCh38
rs781608260 Chr.4: 8605953-8605953 on Build GRCh38
rs761347771 Chr.4: 8606017-8606020 on Build GRCh38
rs758061741 Chr.4: 8606043-8606043 on Build GRCh38
rs202092274 Chr.4: 8606030-8606030 on Build GRCh38
rs751651131 Chr.4: 8605997-8605997 on Build GRCh38
rs200564031 Chr.4: 8605982-8605982 on Build GRCh38
rs761156679 Chr.4: 8606072-8606072 on Build GRCh38
rs546960736 Chr.4: 8605937-8605937 on Build GRCh38
rs192903509 Chr.4: 8605949-8605949 on Build GRCh38
rs528272125 Chr.4: 8605935-8605935 on Build GRCh38
rs368212553 Chr.4: 8606044-8606044 on Build GRCh38
rs764252538 Chr.4: 8605996-8605996 on Build GRCh38
rs555501833 Chr.4: 8606037-8606037 on Build GRCh38
rs774461838 Chr.4: 8606065-8606065 on Build GRCh38
rs115075586 Chr.4: 8606029-8606029 on Build GRCh38
rs749078069 Chr.4: 8606062-8606062 on Build GRCh38
rs756402952 Chr.4: 8606038-8606038 on Build GRCh38
rs374024695 Chr.4: 8605975-8605975 on Build GRCh38
rs777323117 Chr.4: 8606014-8606014 on Build GRCh38
rs779994546 Chr.4: 8606061-8606061 on Build GRCh38
rs770828220 Chr.4: 8605979-8605979 on Build GRCh38
rs946357134 Chr.4: 8605942-8605942 on Build GRCh38
rs765617177 Chr.4: 8605946-8605946 on Build GRCh38
rs753947453 Chr.4: 8605944-8605944 on Build GRCh38
rs981952447 Chr.4: 8605890-8605890 on Build GRCh38
rs146292135 Chr.4: 8606045-8606045 on Build GRCh38
rs752252286 Chr.4: 8605948-8605948 on Build GRCh38
rs568731713 Chr.4: 8606010-8606010 on Build GRCh38
rs763925136 Chr.4: 8606015-8606015 on Build GRCh38
rs533492571 Chr.4: 8605906-8605906 on Build GRCh38
rs767950121 Chr.4: 8605998-8605998 on Build GRCh38
rs747606467 Chr.4: 8605967-8605967 on Build GRCh38
rs781211829 Chr.4: 8606050-8606050 on Build GRCh38
rs796447794 Chr.4: 8605943-8605943 on Build GRCh38
rs565578313 Chr.4: 8605918-8605919 on Build GRCh38
rs769914468 Chr.4: 8606056-8606056 on Build GRCh38
rs200227054 Chr.4: 8606071-8606071 on Build GRCh38
rs900154917 Chr.4: 8605919-8605919 on Build GRCh38
rs768021205 Chr.4: 8606036-8606036 on Build GRCh38
rs757789357 Chr.4: 8606012-8606012 on Build GRCh38
rs770867346 Chr.4: 8606018-8606018 on Build GRCh38
rs759442100 Chr.4: 8605984-8605984 on Build GRCh38
rs139418095 Chr.4: 8606067-8606067 on Build GRCh38
rs971507813 Chr.4: 8606060-8606060 on Build GRCh38
rs776755903 Chr.4: 8606019-8606019 on Build GRCh38
rs778587239 Chr.4: 8605963-8605963 on Build GRCh38
rs368587083 Chr.4: 8605947-8605947 on Build GRCh38
rs761850280 Chr.4: 8606033-8606033 on Build GRCh38
rs201606859 Chr.4: 8605968-8605968 on Build GRCh38
rs140064374 Chr.4: 8606008-8606008 on Build GRCh38
rs746945166 Chr.4: 8605942-8605943 on Build GRCh38
rs368230336 Chr.4: 8605994-8605994 on Build GRCh38
rs551547280 Chr.4: 8605919-8605919 on Build GRCh38
rs765532026 Chr.4: 8605985-8605985 on Build GRCh38
rs372598897 Chr.4: 8606001-8606001 on Build GRCh38
rs768239237 Chr.4: 8605963-8605963 on Build GRCh38
rs113873823 Chr.4: 8606011-8606011 on Build GRCh38
rs768902033 Chr.4: 8606064-8606064 on Build GRCh38
rs562694975 Chr.4: 8605902-8605902 on Build GRCh38
rs557547766 Chr.4: 8606017-8606017 on Build GRCh38
rs374128411 Chr.4: 8605939-8605939 on Build GRCh38
rs543084911 Chr.4: 8605978-8605978 on Build GRCh38
rs745815864 Chr.4: 8606051-8606051 on Build GRCh38
rs769525875 Chr.4: 8606067-8606081 on Build GRCh38
rs768269402 Chr.4: 8605959-8605959 on Build GRCh38
rs766742485 Chr.4: 8606042-8606042 on Build GRCh38
rs758148667 Chr.4: 8605986-8605986 on Build GRCh38
rs769718507 Chr.4: 8606026-8606026 on Build GRCh38

View Full Product Details