Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32879105-32879344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP89
Entrez Gene ID
84902
Gene Name
centrosomal protein 89
Gene Aliases
CCDC123, CEP123
UniGene
Hs.599703
Gene Chromosome Location
Chr.19: 32875925-32972029 on Build GRCh38
Transcripts
NM_032816.4 XM_005259344.3 XM_011527425.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4671738 c.2200G>T p.E734* Chr.19: 32879314-32879314 on Build GRCh38
7170819 c.2270T>G p.L757R Chr.19: 32879244-32879244 on Build GRCh38
4671739 c.2188C>T p.R730* Chr.19: 32879326-32879326 on Build GRCh38
5599286 c.2197C>T p.R733* Chr.19: 32879317-32879317 on Build GRCh38
5038965 c.2315G>T p.C772F Chr.19: 32879199-32879199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757097133 Chr.19: 32879158-32879158 on Build GRCh38
rs780316076 Chr.19: 32879163-32879163 on Build GRCh38
rs779269653 Chr.19: 32879167-32879167 on Build GRCh38
rs532070824 Chr.19: 32879131-32879131 on Build GRCh38
rs776078700 Chr.19: 32879142-32879142 on Build GRCh38
rs775095011 Chr.19: 32879187-32879187 on Build GRCh38
rs199880123 Chr.19: 32879155-32879155 on Build GRCh38
rs770493510 Chr.19: 32879136-32879136 on Build GRCh38
rs377394598 Chr.19: 32879302-32879302 on Build GRCh38
rs991941807 Chr.19: 32879270-32879270 on Build GRCh38
rs769271748 Chr.19: 32879292-32879292 on Build GRCh38
rs139465833 Chr.19: 32879174-32879174 on Build GRCh38
rs745439546 Chr.19: 32879147-32879147 on Build GRCh38
rs763978785 Chr.19: 32879154-32879154 on Build GRCh38
rs769504661 Chr.19: 32879183-32879183 on Build GRCh38
rs201835752 Chr.19: 32879207-32879207 on Build GRCh38
rs201311820 Chr.19: 32879213-32879213 on Build GRCh38
rs569271246 Chr.19: 32879297-32879297 on Build GRCh38
rs537113854 Chr.19: 32879236-32879236 on Build GRCh38
rs531504560 Chr.19: 32879173-32879173 on Build GRCh38
rs983668611 Chr.19: 32879205-32879205 on Build GRCh38
rs908409519 Chr.19: 32879277-32879277 on Build GRCh38
rs113325124 Chr.19: 32879237-32879237 on Build GRCh38
rs761503150 Chr.19: 32879309-32879309 on Build GRCh38
rs146991222 Chr.19: 32879317-32879317 on Build GRCh38
rs774211632 Chr.19: 32879306-32879306 on Build GRCh38
rs202084887 Chr.19: 32879193-32879193 on Build GRCh38
rs377001468 Chr.19: 32879146-32879146 on Build GRCh38
rs141423846 Chr.19: 32879316-32879316 on Build GRCh38
rs762386606 Chr.19: 32879230-32879230 on Build GRCh38
rs373369528 Chr.19: 32879201-32879201 on Build GRCh38
rs745961 Chr.19: 32879164-32879164 on Build GRCh38
rs748498279 Chr.19: 32879169-32879169 on Build GRCh38
rs745424367 Chr.19: 32879281-32879281 on Build GRCh38
rs929169627 Chr.19: 32879196-32879196 on Build GRCh38
rs762805067 Chr.19: 32879150-32879150 on Build GRCh38
rs759032645 Chr.19: 32879179-32879179 on Build GRCh38
rs538660487 Chr.19: 32879252-32879252 on Build GRCh38
rs767599312 Chr.19: 32879314-32879314 on Build GRCh38
rs745960 Chr.19: 32879178-32879178 on Build GRCh38
rs773669899 Chr.19: 32879261-32879261 on Build GRCh38
rs745959 Chr.19: 32879255-32879255 on Build GRCh38
rs755075467 Chr.19: 32879220-32879220 on Build GRCh38
rs774814142 Chr.19: 32879296-32879296 on Build GRCh38
rs759191975 Chr.19: 32879143-32879143 on Build GRCh38
rs778203858 Chr.19: 32879243-32879243 on Build GRCh38
rs772455785 Chr.19: 32879170-32879170 on Build GRCh38
rs905516334 Chr.19: 32879141-32879141 on Build GRCh38
rs773981059 Chr.19: 32879204-32879204 on Build GRCh38

View Full Product Details