Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111457155-111457428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN2
Entrez Gene ID
6311
Gene Name
ataxin 2
Gene Aliases
ASL13, ATX2, SCA2, TNRC13
UniGene
Hs.732512
Gene Chromosome Location
Chr.12: 111452214-111599676 on Build GRCh38
Transcripts
NM_001310121.1 NM_001310123.1 NM_002973.3 NR_132311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251837 c.3508C>A p.P1170T Chr.12: 111457222-111457222 on Build GRCh38
4038728 c.3402G>A p.A1134A Chr.12: 111457328-111457328 on Build GRCh38
1947677 c.3377C>T p.T1126M Chr.12: 111457353-111457353 on Build GRCh38
5456260 c.3371-9T>C p.? Chr.12: 111457368-111457368 on Build GRCh38
4544027 c.3488G>A p.G1163E Chr.12: 111457242-111457242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74882643 Chr.12: 111457232-111457232 on Build GRCh38
rs765497646 Chr.12: 111457334-111457334 on Build GRCh38
rs758751124 Chr.12: 111457352-111457352 on Build GRCh38
rs753915762 Chr.12: 111457319-111457319 on Build GRCh38
rs777327738 Chr.12: 111457391-111457391 on Build GRCh38
rs377369884 Chr.12: 111457365-111457365 on Build GRCh38
rs369927759 Chr.12: 111457393-111457393 on Build GRCh38
rs747639610 Chr.12: 111457238-111457238 on Build GRCh38
rs779534880 Chr.12: 111457356-111457356 on Build GRCh38
rs769294209 Chr.12: 111457386-111457386 on Build GRCh38
rs781011377 Chr.12: 111457382-111457382 on Build GRCh38
rs1009108100 Chr.12: 111457264-111457264 on Build GRCh38
rs12301585 Chr.12: 111457405-111457405 on Build GRCh38
rs373299309 Chr.12: 111457342-111457342 on Build GRCh38
rs964446609 Chr.12: 111457328-111457328 on Build GRCh38
rs1017445924 Chr.12: 111457275-111457275 on Build GRCh38
rs779972223 Chr.12: 111457355-111457355 on Build GRCh38
rs374007476 Chr.12: 111457384-111457384 on Build GRCh38
rs370123294 Chr.12: 111457270-111457270 on Build GRCh38
rs200574928 Chr.12: 111457196-111457196 on Build GRCh38
rs759825988 Chr.12: 111457266-111457266 on Build GRCh38
rs775781088 Chr.12: 111457289-111457289 on Build GRCh38
rs754272313 Chr.12: 111457219-111457219 on Build GRCh38
rs761823722 Chr.12: 111457329-111457329 on Build GRCh38
rs772425521 Chr.12: 111457274-111457274 on Build GRCh38
rs117808303 Chr.12: 111457250-111457250 on Build GRCh38
rs79122454 Chr.12: 111457234-111457234 on Build GRCh38
rs374727874 Chr.12: 111457239-111457239 on Build GRCh38
rs77613739 Chr.12: 111457233-111457233 on Build GRCh38
rs575705515 Chr.12: 111457182-111457182 on Build GRCh38
rs746951692 Chr.12: 111457194-111457194 on Build GRCh38
rs770623367 Chr.12: 111457394-111457394 on Build GRCh38
rs968717790 Chr.12: 111457378-111457378 on Build GRCh38
rs149109789 Chr.12: 111457318-111457318 on Build GRCh38
rs747634376 Chr.12: 111457383-111457383 on Build GRCh38
rs2301622 Chr.12: 111457399-111457399 on Build GRCh38
rs780071979 Chr.12: 111457185-111457185 on Build GRCh38
rs776175107 Chr.12: 111457205-111457205 on Build GRCh38

View Full Product Details