Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74723171-74723442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCF1
Entrez Gene ID
51077
Gene Name
FCF1 rRNA-processing protein
Gene Aliases
Bka, C14orf111, CGI-35, UTP24
UniGene
Hs.579828
Gene Chromosome Location
Chr.14: 74713118-74738620 on Build GRCh38
Transcripts
NM_001318508.1 NM_015962.4 XM_005267730.3 XM_011536815.2 XM_011536816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922496 c.363A>C p.L121L Chr.14: 74723342-74723342 on Build GRCh38
957804 c.299C>A p.P100Q Chr.14: 74723278-74723278 on Build GRCh38
6337576 c.352C>T p.R118* Chr.14: 74723331-74723331 on Build GRCh38
3370059 c.361C>G p.L121V Chr.14: 74723340-74723340 on Build GRCh38
6338458 c.309C>T p.T103T Chr.14: 74723288-74723288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376747411 Chr.14: 74723287-74723287 on Build GRCh38
rs41309250 Chr.14: 74723299-74723299 on Build GRCh38
rs761523720 Chr.14: 74723362-74723362 on Build GRCh38
rs777007341 Chr.14: 74723333-74723333 on Build GRCh38
rs772778377 Chr.14: 74723372-74723372 on Build GRCh38
rs542704795 Chr.14: 74723405-74723406 on Build GRCh38
rs759882637 Chr.14: 74723274-74723274 on Build GRCh38
rs964020844 Chr.14: 74723207-74723207 on Build GRCh38
rs200585479 Chr.14: 74723365-74723365 on Build GRCh38
rs773669558 Chr.14: 74723246-74723246 on Build GRCh38
rs756068598 Chr.14: 74723288-74723288 on Build GRCh38
rs139087378 Chr.14: 74723296-74723296 on Build GRCh38
rs34238686 Chr.14: 74723321-74723321 on Build GRCh38
rs772162581 Chr.14: 74723237-74723237 on Build GRCh38
rs774102719 Chr.14: 74723353-74723353 on Build GRCh38
rs778859623 Chr.14: 74723324-74723325 on Build GRCh38
rs758660817 Chr.14: 74723326-74723326 on Build GRCh38
rs908673607 Chr.14: 74723285-74723285 on Build GRCh38
rs36025752 Chr.14: 74723276-74723276 on Build GRCh38
rs61730183 Chr.14: 74723279-74723279 on Build GRCh38
rs148554557 Chr.14: 74723255-74723256 on Build GRCh38
rs746183205 Chr.14: 74723337-74723337 on Build GRCh38
rs889086200 Chr.14: 74723342-74723342 on Build GRCh38
rs562076830 Chr.14: 74723234-74723234 on Build GRCh38
rs771464162 Chr.14: 74723332-74723332 on Build GRCh38
rs777807521 Chr.14: 74723328-74723328 on Build GRCh38
rs771390230 Chr.14: 74723260-74723260 on Build GRCh38
rs1036497356 Chr.14: 74723323-74723323 on Build GRCh38
rs183412051 Chr.14: 74723250-74723250 on Build GRCh38
rs368964456 Chr.14: 74723344-74723344 on Build GRCh38
rs754745180 Chr.14: 74723308-74723308 on Build GRCh38
rs201408131 Chr.14: 74723331-74723331 on Build GRCh38
rs141609802 Chr.14: 74723309-74723309 on Build GRCh38
rs760111287 Chr.14: 74723393-74723393 on Build GRCh38
rs749099773 Chr.14: 74723228-74723228 on Build GRCh38
rs187365036 Chr.14: 74723264-74723264 on Build GRCh38
rs919468460 Chr.14: 74723329-74723329 on Build GRCh38
rs936394657 Chr.14: 74723399-74723399 on Build GRCh38
rs192295116 Chr.14: 74723281-74723281 on Build GRCh38
rs375092386 Chr.14: 74723230-74723230 on Build GRCh38
rs768920899 Chr.14: 74723229-74723229 on Build GRCh38

View Full Product Details