Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139680914-139681187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXXC5
Entrez Gene ID
51523
Gene Name
CXXC finger protein 5
Gene Aliases
CF5, HSPC195, RINF, WID
UniGene
Hs.189119
Gene Chromosome Location
Chr.5: 139647299-139683885 on Build GRCh38
Transcripts
NM_001317199.1 NM_001317200.1 NM_001317201.1 NM_001317202.1 NM_001317203.1 NM_001317204.1 NM_001317205.1 NM_001317206.1 NM_001317207.1 NM_001317208.1 NM_001317209.1 NM_001317210.1 NM_001317211.1 NM_016463.8 XM_017009573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674291 c.542C>A p.P181Q Chr.5: 139681065-139681065 on Build GRCh38
5684195 c.465A>T p.A155A Chr.5: 139680988-139680988 on Build GRCh38
5737346 c.424G>A p.A142T Chr.5: 139680947-139680947 on Build GRCh38
3850532 c.586G>A p.V196M Chr.5: 139681109-139681109 on Build GRCh38
4675735 c.534G>A p.E178E Chr.5: 139681057-139681057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759082953 Chr.5: 139681066-139681066 on Build GRCh38
rs965831745 Chr.5: 139681142-139681142 on Build GRCh38
rs777041360 Chr.5: 139681120-139681120 on Build GRCh38
rs202150540 Chr.5: 139681117-139681117 on Build GRCh38
rs752404340 Chr.5: 139681149-139681149 on Build GRCh38
rs751188530 Chr.5: 139680969-139680969 on Build GRCh38
rs772377109 Chr.5: 139681015-139681015 on Build GRCh38
rs769024620 Chr.5: 139681119-139681119 on Build GRCh38
rs755975384 Chr.5: 139681081-139681081 on Build GRCh38
rs748020882 Chr.5: 139680981-139680981 on Build GRCh38
rs775697390 Chr.5: 139681017-139681017 on Build GRCh38
rs544140536 Chr.5: 139680940-139680940 on Build GRCh38
rs780355584 Chr.5: 139681110-139681110 on Build GRCh38
rs764545803 Chr.5: 139681023-139681023 on Build GRCh38
rs1051486370 Chr.5: 139681065-139681065 on Build GRCh38
rs867525029 Chr.5: 139681028-139681028 on Build GRCh38
rs746008707 Chr.5: 139681101-139681101 on Build GRCh38
rs760460579 Chr.5: 139681152-139681152 on Build GRCh38
rs751152936 Chr.5: 139681058-139681058 on Build GRCh38
rs755954309 Chr.5: 139680991-139680991 on Build GRCh38
rs773682660 Chr.5: 139681127-139681127 on Build GRCh38
rs762395670 Chr.5: 139681044-139681044 on Build GRCh38
rs749310451 Chr.5: 139680999-139680999 on Build GRCh38
rs370610029 Chr.5: 139681076-139681076 on Build GRCh38
rs763974365 Chr.5: 139681160-139681160 on Build GRCh38
rs199510687 Chr.5: 139680992-139680992 on Build GRCh38
rs772136553 Chr.5: 139681109-139681109 on Build GRCh38
rs914458547 Chr.5: 139680985-139680985 on Build GRCh38
rs545131681 Chr.5: 139681043-139681043 on Build GRCh38
rs771147937 Chr.5: 139681005-139681005 on Build GRCh38
rs113958458 Chr.5: 139680972-139680972 on Build GRCh38
rs777792508 Chr.5: 139681082-139681082 on Build GRCh38
rs754719545 Chr.5: 139680971-139680971 on Build GRCh38
rs759113157 Chr.5: 139681132-139681132 on Build GRCh38
rs753812369 Chr.5: 139681086-139681086 on Build GRCh38
rs999128990 Chr.5: 139681072-139681072 on Build GRCh38
rs746071745 Chr.5: 139681009-139681009 on Build GRCh38
rs183852682 Chr.5: 139681046-139681046 on Build GRCh38
rs780847835 Chr.5: 139680979-139680979 on Build GRCh38
rs767217574 Chr.5: 139681075-139681075 on Build GRCh38
rs373625100 Chr.5: 139681006-139681006 on Build GRCh38
rs528724719 Chr.5: 139681091-139681091 on Build GRCh38
rs1045910763 Chr.5: 139681060-139681060 on Build GRCh38
rs909915231 Chr.5: 139680995-139680995 on Build GRCh38
rs778826979 Chr.5: 139681099-139681099 on Build GRCh38
rs757935649 Chr.5: 139680939-139680939 on Build GRCh38
rs373699733 Chr.5: 139681141-139681141 on Build GRCh38
rs1052970340 Chr.5: 139680938-139680938 on Build GRCh38
rs371784240 Chr.5: 139681021-139681021 on Build GRCh38
rs373740936 Chr.5: 139681045-139681045 on Build GRCh38

View Full Product Details