Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96114663-96114902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA2B
Entrez Gene ID
151
Gene Name
adrenoceptor alpha 2B
Gene Aliases
ADRA2L1, ADRA2RL1, ADRARL1, ALPHA2BAR, FAME2, alpha-2BAR
UniGene
Hs.247686
Gene Chromosome Location
Chr.2: 96112875-96116245 on Build GRCh38
Transcripts
NM_000682.6

SNP Detail

SNP ID Chromosome Location
rs562532135 Chr.2: 96114850-96114850 on Build GRCh38
rs75133239 Chr.2: 96114765-96114765 on Build GRCh38
rs928347207 Chr.2: 96114864-96114864 on Build GRCh38
rs528195928 Chr.2: 96114787-96114787 on Build GRCh38
rs766575378 Chr.2: 96114793-96114793 on Build GRCh38
rs369918003 Chr.2: 96114844-96114844 on Build GRCh38
rs367910193 Chr.2: 96114805-96114805 on Build GRCh38
rs763020664 Chr.2: 96114872-96114872 on Build GRCh38
rs34894270 Chr.2: 96114792-96114792 on Build GRCh38
rs765425973 Chr.2: 96114856-96114856 on Build GRCh38
rs755499239 Chr.2: 96114798-96114799 on Build GRCh38
rs200055567 Chr.2: 96114807-96114807 on Build GRCh38
rs749269879 Chr.2: 96114786-96114786 on Build GRCh38
rs556484226 Chr.2: 96114754-96114754 on Build GRCh38
rs759782711 Chr.2: 96114849-96114849 on Build GRCh38
rs753214024 Chr.2: 96114755-96114755 on Build GRCh38
rs368428483 Chr.2: 96114758-96114758 on Build GRCh38
rs906851956 Chr.2: 96114733-96114733 on Build GRCh38
rs770552082 Chr.2: 96114764-96114764 on Build GRCh38
rs539857970 Chr.2: 96114819-96114819 on Build GRCh38
rs1041898466 Chr.2: 96114714-96114714 on Build GRCh38
rs778137476 Chr.2: 96114882-96114883 on Build GRCh38
rs775092649 Chr.2: 96114773-96114773 on Build GRCh38
rs768772295 Chr.2: 96114839-96114839 on Build GRCh38
rs376416256 Chr.2: 96114817-96114817 on Build GRCh38
rs186085130 Chr.2: 96114795-96114795 on Build GRCh38
rs751939546 Chr.2: 96114815-96114815 on Build GRCh38
rs553609439 Chr.2: 96114823-96114823 on Build GRCh38
rs752731456 Chr.2: 96114806-96114806 on Build GRCh38
rs755789092 Chr.2: 96114827-96114827 on Build GRCh38
rs764094629 Chr.2: 96114747-96114747 on Build GRCh38
rs892981408 Chr.2: 96114708-96114708 on Build GRCh38
rs781601166 Chr.2: 96114846-96114846 on Build GRCh38
rs577064249 Chr.2: 96114838-96114838 on Build GRCh38
rs765321482 Chr.2: 96114799-96114799 on Build GRCh38
rs35355631 Chr.2: 96114756-96114756 on Build GRCh38
rs781353874 Chr.2: 96114822-96114822 on Build GRCh38
rs764017593 Chr.2: 96114883-96114883 on Build GRCh38
rs771910033 Chr.2: 96114846-96114846 on Build GRCh38
rs546124409 Chr.2: 96114847-96114847 on Build GRCh38
rs371368208 Chr.2: 96114750-96114750 on Build GRCh38
rs748485032 Chr.2: 96114851-96114853 on Build GRCh38
rs770108103 Chr.2: 96114861-96114861 on Build GRCh38
rs779749286 Chr.2: 96114837-96114837 on Build GRCh38
rs372042632 Chr.2: 96114759-96114759 on Build GRCh38
rs29000570 Chr.2: 96114799-96114800 on Build GRCh38
rs769442805 Chr.2: 96114768-96114768 on Build GRCh38
rs1006000772 Chr.2: 96114692-96114692 on Build GRCh38
rs35801656 Chr.2: 96114821-96114822 on Build GRCh38
rs931866175 Chr.2: 96114818-96114818 on Build GRCh38
rs945793647 Chr.2: 96114712-96114712 on Build GRCh38
rs775606736 Chr.2: 96114857-96114857 on Build GRCh38

View Full Product Details