Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140562056-140562305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB3
Entrez Gene ID
10307
Gene Name
amyloid beta precursor protein binding family B member 3
Gene Aliases
FE65L2, SRA
UniGene
Hs.529449
Gene Chromosome Location
Chr.5: 140558268-140564604 on Build GRCh38
Transcripts
NM_006051.3 NM_133172.2 NM_133173.2 NM_133174.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3850670 c.607G>A p.V203M Chr.5: 140562119-140562119 on Build GRCh38
3850671 c.533C>T p.A178V Chr.5: 140562193-140562193 on Build GRCh38
1754016 c.610G>C p.G204R Chr.5: 140562116-140562116 on Build GRCh38
81998 c.561C>T p.H187H Chr.5: 140562165-140562165 on Build GRCh38
3975195 c.578A>T p.Q193L Chr.5: 140562148-140562148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147739115 Chr.5: 140562207-140562207 on Build GRCh38
rs145695941 Chr.5: 140562175-140562175 on Build GRCh38
rs201714439 Chr.5: 140562100-140562100 on Build GRCh38
rs140070711 Chr.5: 140562130-140562130 on Build GRCh38
rs148789210 Chr.5: 140562220-140562220 on Build GRCh38
rs766301321 Chr.5: 140562259-140562259 on Build GRCh38
rs920129674 Chr.5: 140562076-140562076 on Build GRCh38
rs199700470 Chr.5: 140562090-140562090 on Build GRCh38
rs375776763 Chr.5: 140562277-140562277 on Build GRCh38
rs979022363 Chr.5: 140562097-140562097 on Build GRCh38
rs772877296 Chr.5: 140562243-140562243 on Build GRCh38
rs777291861 Chr.5: 140562133-140562133 on Build GRCh38
rs750242454 Chr.5: 140562127-140562127 on Build GRCh38
rs781456479 Chr.5: 140562156-140562156 on Build GRCh38
rs7734425 Chr.5: 140562275-140562275 on Build GRCh38
rs182314255 Chr.5: 140562270-140562270 on Build GRCh38
rs145227435 Chr.5: 140562215-140562215 on Build GRCh38
rs764874015 Chr.5: 140562122-140562122 on Build GRCh38
rs200825219 Chr.5: 140562101-140562101 on Build GRCh38
rs190071068 Chr.5: 140562079-140562079 on Build GRCh38
rs372228585 Chr.5: 140562120-140562120 on Build GRCh38
rs766488336 Chr.5: 140562148-140562148 on Build GRCh38
rs376576611 Chr.5: 140562228-140562228 on Build GRCh38
rs776216532 Chr.5: 140562119-140562119 on Build GRCh38
rs779443597 Chr.5: 140562209-140562209 on Build GRCh38
rs947496755 Chr.5: 140562236-140562236 on Build GRCh38
rs755063242 Chr.5: 140562153-140562153 on Build GRCh38
rs749729906 Chr.5: 140562186-140562186 on Build GRCh38
rs1047891225 Chr.5: 140562245-140562245 on Build GRCh38
rs769543936 Chr.5: 140562238-140562238 on Build GRCh38
rs955849006 Chr.5: 140562102-140562102 on Build GRCh38
rs759028787 Chr.5: 140562268-140562269 on Build GRCh38
rs917152798 Chr.5: 140562126-140562126 on Build GRCh38
rs368256750 Chr.5: 140562254-140562254 on Build GRCh38
rs544391182 Chr.5: 140562128-140562128 on Build GRCh38
rs751655734 Chr.5: 140562149-140562149 on Build GRCh38
rs768114183 Chr.5: 140562219-140562219 on Build GRCh38
rs763711691 Chr.5: 140562118-140562118 on Build GRCh38
rs752904001 Chr.5: 140562170-140562170 on Build GRCh38
rs777988103 Chr.5: 140562183-140562183 on Build GRCh38
rs756086342 Chr.5: 140562110-140562110 on Build GRCh38

View Full Product Details