Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159782308-159782537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP23
Entrez Gene ID
54935
Gene Name
dual specificity phosphatase 23
Gene Aliases
DUSP25, LDP-3, MOSP, VHZ
UniGene
Hs.425801
Gene Chromosome Location
Chr.1: 159780946-159782546 on Build GRCh38
Transcripts
NM_001319658.1 NM_001319659.1 NM_017823.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898057 c.443G>A p.R148Q Chr.1: 159782328-159782328 on Build GRCh38
6508017 c.447G>A p.T149T Chr.1: 159782332-159782332 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756645706 Chr.1: 159782379-159782379 on Build GRCh38
rs781663031 Chr.1: 159782360-159782360 on Build GRCh38
rs151195156 Chr.1: 159782465-159782465 on Build GRCh38
rs771854825 Chr.1: 159782470-159782470 on Build GRCh38
rs755293751 Chr.1: 159782354-159782354 on Build GRCh38
rs761649173 Chr.1: 159782409-159782409 on Build GRCh38
rs753084244 Chr.1: 159782375-159782375 on Build GRCh38
rs1019909355 Chr.1: 159782395-159782395 on Build GRCh38
rs1129932 Chr.1: 159782356-159782356 on Build GRCh38
rs549990995 Chr.1: 159782387-159782387 on Build GRCh38
rs989802360 Chr.1: 159782364-159782372 on Build GRCh38
rs749398895 Chr.1: 159782382-159782382 on Build GRCh38
rs535617185 Chr.1: 159782499-159782499 on Build GRCh38
rs779248955 Chr.1: 159782390-159782390 on Build GRCh38
rs901236775 Chr.1: 159782462-159782462 on Build GRCh38
rs1007122679 Chr.1: 159782389-159782389 on Build GRCh38
rs1042427137 Chr.1: 159782432-159782432 on Build GRCh38
rs764846450 Chr.1: 159782343-159782343 on Build GRCh38
rs934046874 Chr.1: 159782483-159782483 on Build GRCh38
rs777958090 Chr.1: 159782381-159782381 on Build GRCh38

View Full Product Details