Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67646737-67646972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACY3
Entrez Gene ID
91703
Gene Name
aminoacylase 3
Gene Aliases
ACY-3, HCBP1
UniGene
Hs.126265
Gene Chromosome Location
Chr.11: 67642551-67650829 on Build GRCh38
Transcripts
NM_080658.1 XM_017018549.1 XM_017018550.1 XM_017018551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3356243 c.227G>A p.S76N Chr.11: 67646817-67646817 on Build GRCh38
4659239 c.169G>A p.A57T Chr.11: 67646875-67646875 on Build GRCh38
7155782 c.230T>G p.F77C Chr.11: 67646814-67646814 on Build GRCh38
5641749 c.91G>T p.A31S Chr.11: 67646953-67646953 on Build GRCh38
4645796 c.138C>T p.F46F Chr.11: 67646906-67646906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920295276 Chr.11: 67646897-67646897 on Build GRCh38
rs779671699 Chr.11: 67646950-67646950 on Build GRCh38
rs890212929 Chr.11: 67646939-67646939 on Build GRCh38
rs748420083 Chr.11: 67646849-67646849 on Build GRCh38
rs778074437 Chr.11: 67646852-67646852 on Build GRCh38
rs374601574 Chr.11: 67646782-67646782 on Build GRCh38
rs868266214 Chr.11: 67646953-67646953 on Build GRCh38
rs778074987 Chr.11: 67646924-67646924 on Build GRCh38
rs755097737 Chr.11: 67646787-67646787 on Build GRCh38
rs140848754 Chr.11: 67646879-67646879 on Build GRCh38
rs746428118 Chr.11: 67646934-67646934 on Build GRCh38
rs968727262 Chr.11: 67646841-67646841 on Build GRCh38
rs185851624 Chr.11: 67646792-67646792 on Build GRCh38
rs369691782 Chr.11: 67646856-67646856 on Build GRCh38
rs779948415 Chr.11: 67646776-67646776 on Build GRCh38
rs759096432 Chr.11: 67646803-67646803 on Build GRCh38
rs758324349 Chr.11: 67646765-67646765 on Build GRCh38
rs145934175 Chr.11: 67646880-67646880 on Build GRCh38
rs948444 Chr.11: 67646775-67646775 on Build GRCh38
rs752205792 Chr.11: 67646842-67646842 on Build GRCh38
rs546417694 Chr.11: 67646764-67646764 on Build GRCh38
rs149075333 Chr.11: 67646772-67646772 on Build GRCh38
rs533005301 Chr.11: 67646869-67646869 on Build GRCh38
rs773950556 Chr.11: 67646882-67646882 on Build GRCh38
rs148918245 Chr.11: 67646809-67646809 on Build GRCh38
rs759588553 Chr.11: 67646903-67646903 on Build GRCh38
rs747428964 Chr.11: 67646795-67646795 on Build GRCh38
rs757103037 Chr.11: 67646831-67646831 on Build GRCh38
rs764981933 Chr.11: 67646758-67646758 on Build GRCh38
rs752801343 Chr.11: 67646911-67646911 on Build GRCh38
rs769467221 Chr.11: 67646866-67646866 on Build GRCh38
rs758545304 Chr.11: 67646912-67646912 on Build GRCh38
rs138978537 Chr.11: 67646881-67646881 on Build GRCh38
rs928996990 Chr.11: 67646833-67646833 on Build GRCh38
rs753685871 Chr.11: 67646873-67646874 on Build GRCh38
rs758644522 Chr.11: 67646851-67646851 on Build GRCh38
rs745363368 Chr.11: 67646860-67646860 on Build GRCh38
rs981745209 Chr.11: 67646883-67646883 on Build GRCh38
rs771503467 Chr.11: 67646798-67646798 on Build GRCh38
rs753850557 Chr.11: 67646839-67646839 on Build GRCh38
rs775346579 Chr.11: 67646807-67646807 on Build GRCh38
rs370668700 Chr.11: 67646929-67646929 on Build GRCh38
rs536521467 Chr.11: 67646805-67646805 on Build GRCh38
rs764029118 Chr.11: 67646819-67646819 on Build GRCh38
rs543980460 Chr.11: 67646857-67646857 on Build GRCh38
rs373688915 Chr.11: 67646887-67646887 on Build GRCh38
rs767503729 Chr.11: 67646832-67646832 on Build GRCh38
rs764121194 Chr.11: 67646771-67646771 on Build GRCh38
rs1008408230 Chr.11: 67646781-67646781 on Build GRCh38
rs751392179 Chr.11: 67646828-67646828 on Build GRCh38
rs770448858 Chr.11: 67646949-67646949 on Build GRCh38
rs377603005 Chr.11: 67646927-67646927 on Build GRCh38
rs767350912 Chr.11: 67646890-67646890 on Build GRCh38
rs750414025 Chr.11: 67646899-67646899 on Build GRCh38
rs376968070 Chr.11: 67646842-67646842 on Build GRCh38
rs143187416 Chr.11: 67646909-67646909 on Build GRCh38
rs775932634 Chr.11: 67646802-67646802 on Build GRCh38
rs143717535 Chr.11: 67646848-67646848 on Build GRCh38
rs151057744 Chr.11: 67646859-67646859 on Build GRCh38
rs554880765 Chr.11: 67646867-67646867 on Build GRCh38

View Full Product Details