Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84425684-84425935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCSER2
Entrez Gene ID
54462
Gene Name
coiled-coil serine rich protein 2
Gene Aliases
FAM190B, Gcap14, KIAA1128, bA486O22.1
UniGene
Hs.461988
Gene Chromosome Location
Chr.10: 84328558-84518521 on Build GRCh38
Transcripts
NM_001284240.1 NM_001284242.1 NM_001284243.1 NM_018999.3 XM_005269905.3 XM_005269906.3 XM_005269908.3 XM_006717894.1 XM_006717895.1 XM_011539870.2 XM_011539871.2 XM_011539872.2 XM_011539873.1 XM_017016340.1 XM_017016341.1 XM_017016342.1 XM_017016343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466023 c.1775G>T p.W592L Chr.10: 84425800-84425800 on Build GRCh38
3415292 c.1727G>A p.R576H Chr.10: 84425752-84425752 on Build GRCh38
3807855 c.1826A>C p.H609P Chr.10: 84425851-84425851 on Build GRCh38
6452053 c.1726C>T p.R576C Chr.10: 84425751-84425751 on Build GRCh38
5979927 c.1784C>T p.P595L Chr.10: 84425809-84425809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867829544 Chr.10: 84425871-84425871 on Build GRCh38
rs764575141 Chr.10: 84425722-84425722 on Build GRCh38
rs1033141671 Chr.10: 84425761-84425761 on Build GRCh38
rs751019604 Chr.10: 84425853-84425853 on Build GRCh38
rs144839234 Chr.10: 84425779-84425779 on Build GRCh38
rs755677090 Chr.10: 84425888-84425888 on Build GRCh38
rs780546894 Chr.10: 84425778-84425778 on Build GRCh38
rs780899399 Chr.10: 84425869-84425869 on Build GRCh38
rs765396988 Chr.10: 84425852-84425852 on Build GRCh38
rs145429708 Chr.10: 84425835-84425835 on Build GRCh38
rs776652929 Chr.10: 84425836-84425836 on Build GRCh38
rs370948089 Chr.10: 84425752-84425752 on Build GRCh38
rs773073885 Chr.10: 84425718-84425718 on Build GRCh38
rs138754897 Chr.10: 84425788-84425788 on Build GRCh38
rs770762144 Chr.10: 84425811-84425811 on Build GRCh38
rs185632970 Chr.10: 84425863-84425863 on Build GRCh38
rs746615001 Chr.10: 84425902-84425903 on Build GRCh38
rs763526687 Chr.10: 84425856-84425856 on Build GRCh38
rs758894670 Chr.10: 84425777-84425777 on Build GRCh38
rs551355079 Chr.10: 84425769-84425769 on Build GRCh38
rs751971470 Chr.10: 84425860-84425860 on Build GRCh38
rs760929089 Chr.10: 84425743-84425743 on Build GRCh38
rs369294950 Chr.10: 84425906-84425906 on Build GRCh38
rs374305577 Chr.10: 84425799-84425799 on Build GRCh38
rs958393758 Chr.10: 84425765-84425765 on Build GRCh38
rs141796865 Chr.10: 84425793-84425793 on Build GRCh38
rs762738070 Chr.10: 84425724-84425724 on Build GRCh38
rs757781154 Chr.10: 84425723-84425724 on Build GRCh38
rs778657495 Chr.10: 84425900-84425900 on Build GRCh38
rs1336202 Chr.10: 84425898-84425898 on Build GRCh38
rs968168104 Chr.10: 84425892-84425892 on Build GRCh38
rs372819578 Chr.10: 84425849-84425849 on Build GRCh38
rs759203869 Chr.10: 84425834-84425834 on Build GRCh38
rs527341764 Chr.10: 84425828-84425828 on Build GRCh38
rs752285093 Chr.10: 84425882-84425882 on Build GRCh38
rs766955590 Chr.10: 84425859-84425859 on Build GRCh38
rs754303811 Chr.10: 84425807-84425807 on Build GRCh38
rs775745511 Chr.10: 84425742-84425742 on Build GRCh38
rs533268688 Chr.10: 84425744-84425744 on Build GRCh38
rs905296317 Chr.10: 84425763-84425763 on Build GRCh38
rs368729802 Chr.10: 84425809-84425809 on Build GRCh38
rs149112423 Chr.10: 84425751-84425751 on Build GRCh38
rs777355436 Chr.10: 84425749-84425757 on Build GRCh38
rs367802673 Chr.10: 84425893-84425893 on Build GRCh38
rs770707093 Chr.10: 84425733-84425733 on Build GRCh38

View Full Product Details