Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52860867-52861134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
UniGene
Hs.509343
Gene Chromosome Location
Chr.14: 52857271-52951192 on Build GRCh38
Transcripts
NM_001134999.1 NM_001135000.1 NM_006832.2 XM_005267285.2 XM_006720008.2 XM_006720009.2 XM_006720010.2

SNP Detail

SNP ID Chromosome Location
rs546083775 Chr.14: 52861038-52861038 on Build GRCh38
rs540304579 Chr.14: 52860981-52860981 on Build GRCh38
rs560845434 Chr.14: 52861086-52861086 on Build GRCh38
rs764887588 Chr.14: 52861073-52861073 on Build GRCh38
rs753890238 Chr.14: 52860951-52860951 on Build GRCh38
rs185865262 Chr.14: 52861084-52861084 on Build GRCh38
rs761290096 Chr.14: 52861049-52861049 on Build GRCh38
rs964362296 Chr.14: 52861031-52861031 on Build GRCh38
rs555146665 Chr.14: 52860948-52860948 on Build GRCh38
rs189588276 Chr.14: 52861025-52861025 on Build GRCh38
rs757245445 Chr.14: 52861048-52861048 on Build GRCh38
rs539338784 Chr.14: 52860899-52860899 on Build GRCh38
rs566455066 Chr.14: 52861075-52861075 on Build GRCh38
rs1026193694 Chr.14: 52860931-52860931 on Build GRCh38
rs747763991 Chr.14: 52861013-52861013 on Build GRCh38
rs772553206 Chr.14: 52861043-52861043 on Build GRCh38
rs974395334 Chr.14: 52861000-52861000 on Build GRCh38
rs577037564 Chr.14: 52861032-52861032 on Build GRCh38
rs773659932 Chr.14: 52861075-52861076 on Build GRCh38
rs780890362 Chr.14: 52860998-52860998 on Build GRCh38
rs768687328 Chr.14: 52861050-52861050 on Build GRCh38
rs1000191870 Chr.14: 52860993-52860993 on Build GRCh38
rs780785228 Chr.14: 52861038-52861039 on Build GRCh38
rs761066397 Chr.14: 52860978-52860979 on Build GRCh38
rs761969897 Chr.14: 52861059-52861059 on Build GRCh38
rs1023307561 Chr.14: 52860897-52860897 on Build GRCh38
rs547857008 Chr.14: 52861106-52861106 on Build GRCh38
rs768216920 Chr.14: 52860971-52860974 on Build GRCh38
rs984569154 Chr.14: 52860950-52860950 on Build GRCh38
rs746221256 Chr.14: 52861040-52861040 on Build GRCh38
rs544897822 Chr.14: 52860909-52860909 on Build GRCh38
rs572258689 Chr.14: 52860959-52860959 on Build GRCh38
rs550551939 Chr.14: 52861107-52861107 on Build GRCh38
rs763289720 Chr.14: 52861076-52861076 on Build GRCh38
rs565056386 Chr.14: 52860932-52860932 on Build GRCh38
rs937092471 Chr.14: 52860895-52860895 on Build GRCh38
rs374898552 Chr.14: 52860965-52860965 on Build GRCh38
rs776719871 Chr.14: 52861051-52861051 on Build GRCh38
rs895192860 Chr.14: 52861095-52861095 on Build GRCh38
rs748555323 Chr.14: 52861054-52861070 on Build GRCh38
rs1015995552 Chr.14: 52860954-52860954 on Build GRCh38
rs139077133 Chr.14: 52860910-52860910 on Build GRCh38
rs193081218 Chr.14: 52861076-52861076 on Build GRCh38

View Full Product Details