Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47568599-47568864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAF
Entrez Gene ID
369
Gene Name
A-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
A-RAF, ARAF1, PKS2, RAFA1
UniGene
Hs.446641
Gene Chromosome Location
Chr.X: 47561100-47571921 on Build GRCh38
Transcripts
NM_001256196.1 NM_001654.4 XM_006724529.2 XM_011543906.2 XM_011543908.2 XM_011543909.2

SNP Detail

SNP ID Chromosome Location
rs145056294 Chr.X: 47568765-47568765 on Build GRCh38
rs906768787 Chr.X: 47568784-47568784 on Build GRCh38
rs778119620 Chr.X: 47568789-47568789 on Build GRCh38
rs886336538 Chr.X: 47568814-47568814 on Build GRCh38
rs755308260 Chr.X: 47568764-47568764 on Build GRCh38
rs898619104 Chr.X: 47568710-47568710 on Build GRCh38
rs764255925 Chr.X: 47568840-47568840 on Build GRCh38
rs771326103 Chr.X: 47568676-47568676 on Build GRCh38
rs756741810 Chr.X: 47568725-47568725 on Build GRCh38
rs774700067 Chr.X: 47568678-47568678 on Build GRCh38
rs774360890 Chr.X: 47568659-47568659 on Build GRCh38
rs367956581 Chr.X: 47568844-47568844 on Build GRCh38
rs767276942 Chr.X: 47568697-47568697 on Build GRCh38
rs756684682 Chr.X: 47568769-47568769 on Build GRCh38
rs765643325 Chr.X: 47568769-47568769 on Build GRCh38
rs760613124 Chr.X: 47568714-47568714 on Build GRCh38
rs759903646 Chr.X: 47568764-47568766 on Build GRCh38
rs771127790 Chr.X: 47568803-47568803 on Build GRCh38
rs759472903 Chr.X: 47568688-47568688 on Build GRCh38
rs763548006 Chr.X: 47568723-47568723 on Build GRCh38
rs753171944 Chr.X: 47568766-47568766 on Build GRCh38
rs750008187 Chr.X: 47568748-47568748 on Build GRCh38
rs749435690 Chr.X: 47568801-47568801 on Build GRCh38
rs754803899 Chr.X: 47568839-47568839 on Build GRCh38
rs942773911 Chr.X: 47568658-47568658 on Build GRCh38
rs759554784 Chr.X: 47568724-47568724 on Build GRCh38
rs779283787 Chr.X: 47568826-47568826 on Build GRCh38
rs753607536 Chr.X: 47568730-47568730 on Build GRCh38
rs184888793 Chr.X: 47568698-47568698 on Build GRCh38
rs772190588 Chr.X: 47568763-47568763 on Build GRCh38
rs1038871440 Chr.X: 47568701-47568701 on Build GRCh38

View Full Product Details