Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34516361-34516622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAPP
Entrez Gene ID
55837
Gene Name
E2F associated phosphoprotein
Gene Aliases
BM036, C14orf11
UniGene
Hs.433269
Gene Chromosome Location
Chr.14: 34515929-34539737 on Build GRCh38
Transcripts
NM_001318916.1 NM_018453.3 XM_011536958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4974284 c.699G>A p.K233K Chr.14: 34516469-34516469 on Build GRCh38
3671904 c.767C>A p.P256Q Chr.14: 34516401-34516401 on Build GRCh38
5015221 c.682A>G p.K228E Chr.14: 34516486-34516486 on Build GRCh38
377460 c.804C>G p.V268V Chr.14: 34516364-34516364 on Build GRCh38
3814720 c.646G>A p.E216K Chr.14: 34516522-34516522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764507994 Chr.14: 34516455-34516455 on Build GRCh38
rs368875293 Chr.14: 34516507-34516507 on Build GRCh38
rs376641920 Chr.14: 34516438-34516438 on Build GRCh38
rs765628827 Chr.14: 34516554-34516554 on Build GRCh38
rs762573321 Chr.14: 34516577-34516577 on Build GRCh38
rs754650838 Chr.14: 34516489-34516489 on Build GRCh38
rs769783651 Chr.14: 34516407-34516407 on Build GRCh38
rs748089841 Chr.14: 34516403-34516403 on Build GRCh38
rs775493364 Chr.14: 34516537-34516537 on Build GRCh38
rs760947629 Chr.14: 34516539-34516539 on Build GRCh38
rs374807491 Chr.14: 34516481-34516481 on Build GRCh38
rs7797 Chr.14: 34516439-34516439 on Build GRCh38
rs765675960 Chr.14: 34516469-34516469 on Build GRCh38
rs577244593 Chr.14: 34516536-34516536 on Build GRCh38
rs777816945 Chr.14: 34516514-34516514 on Build GRCh38
rs779887762 Chr.14: 34516427-34516427 on Build GRCh38
rs765907848 Chr.14: 34516565-34516565 on Build GRCh38
rs939639524 Chr.14: 34516453-34516453 on Build GRCh38
rs749225812 Chr.14: 34516519-34516519 on Build GRCh38
rs372207592 Chr.14: 34516534-34516534 on Build GRCh38
rs375175242 Chr.14: 34516550-34516550 on Build GRCh38
rs781189446 Chr.14: 34516401-34516401 on Build GRCh38
rs767227277 Chr.14: 34516589-34516589 on Build GRCh38
rs780992512 Chr.14: 34516499-34516499 on Build GRCh38
rs76176562 Chr.14: 34516500-34516500 on Build GRCh38
rs374367804 Chr.14: 34516393-34516394 on Build GRCh38
rs763427468 Chr.14: 34516564-34516564 on Build GRCh38
rs762172753 Chr.14: 34516461-34516461 on Build GRCh38
rs765777624 Chr.14: 34516535-34516535 on Build GRCh38
rs768694272 Chr.14: 34516544-34516544 on Build GRCh38
rs17856037 Chr.14: 34516452-34516452 on Build GRCh38
rs201504462 Chr.14: 34516460-34516460 on Build GRCh38
rs750958144 Chr.14: 34516473-34516473 on Build GRCh38
rs775754345 Chr.14: 34516449-34516449 on Build GRCh38
rs368178093 Chr.14: 34516557-34516557 on Build GRCh38
rs776853551 Chr.14: 34516546-34516546 on Build GRCh38
rs149096582 Chr.14: 34516479-34516479 on Build GRCh38
rs944987827 Chr.14: 34516588-34516588 on Build GRCh38
rs993981780 Chr.14: 34516508-34516508 on Build GRCh38
rs779479761 Chr.14: 34516480-34516480 on Build GRCh38
rs773293643 Chr.14: 34516430-34516430 on Build GRCh38
rs768475108 Chr.14: 34516383-34516383 on Build GRCh38

View Full Product Details