Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226597278-226597535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STUM
Entrez Gene ID
375057
Gene Name
stum, mechanosensory transduction mediator homolog
Gene Aliases
C1orf95
UniGene
Hs.116827
Gene Chromosome Location
Chr.1: 226548238-226609214 on Build GRCh38
Transcripts
NM_001003665.3 XM_011544183.2

SNP Detail

SNP ID Chromosome Location
rs921447445 Chr.1: 226597391-226597391 on Build GRCh38
rs775286972 Chr.1: 226597411-226597411 on Build GRCh38
rs562453299 Chr.1: 226597416-226597416 on Build GRCh38
rs930959039 Chr.1: 226597322-226597323 on Build GRCh38
rs73098850 Chr.1: 226597401-226597401 on Build GRCh38
rs756147257 Chr.1: 226597504-226597504 on Build GRCh38
rs573600243 Chr.1: 226597392-226597392 on Build GRCh38
rs1022853685 Chr.1: 226597456-226597456 on Build GRCh38
rs558540999 Chr.1: 226597429-226597429 on Build GRCh38
rs751314135 Chr.1: 226597309-226597309 on Build GRCh38
rs757495538 Chr.1: 226597364-226597364 on Build GRCh38
rs564697484 Chr.1: 226597512-226597512 on Build GRCh38
rs940037510 Chr.1: 226597410-226597410 on Build GRCh38
rs771802533 Chr.1: 226597484-226597484 on Build GRCh38
rs1012851601 Chr.1: 226597433-226597433 on Build GRCh38
rs1039911181 Chr.1: 226597323-226597323 on Build GRCh38
rs150279263 Chr.1: 226597409-226597409 on Build GRCh38
rs770939137 Chr.1: 226597362-226597362 on Build GRCh38
rs367854145 Chr.1: 226597475-226597475 on Build GRCh38
rs138927745 Chr.1: 226597471-226597471 on Build GRCh38

View Full Product Details