Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95042701-95042974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C8
Entrez Gene ID
1558
Gene Name
cytochrome P450 family 2 subfamily C member 8
Gene Aliases
CPC8, CYPIIC8, MP-12/MP-20
UniGene
Hs.709188
Gene Chromosome Location
Chr.10: 95036772-95069512 on Build GRCh38
Transcripts
NM_000770.3 NM_001198853.1 NM_001198854.1 NM_001198855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6130751 c.838G>T p.V280L Chr.10: 95042943-95042943 on Build GRCh38
7328981 c.850G>C p.V284L Chr.10: 95042931-95042931 on Build GRCh38
5020207 c.838C>T p.P280S Chr.10: 95042895-95042895 on Build GRCh38
6130750 c.790G>T p.V264L Chr.10: 95042943-95042943 on Build GRCh38
6189505 c.857C>G p.T286S Chr.10: 95042924-95042924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199581268 Chr.10: 95042843-95042843 on Build GRCh38
rs761663173 Chr.10: 95042763-95042763 on Build GRCh38
rs139182192 Chr.10: 95042822-95042822 on Build GRCh38
rs772497816 Chr.10: 95042952-95042952 on Build GRCh38
rs746442257 Chr.10: 95042951-95042951 on Build GRCh38
rs775358076 Chr.10: 95042806-95042806 on Build GRCh38
rs760009863 Chr.10: 95042884-95042884 on Build GRCh38
rs375271607 Chr.10: 95042895-95042895 on Build GRCh38
rs369479818 Chr.10: 95042947-95042947 on Build GRCh38
rs538552836 Chr.10: 95042766-95042766 on Build GRCh38
rs2275620 Chr.10: 95042841-95042841 on Build GRCh38
rs751169688 Chr.10: 95042911-95042911 on Build GRCh38
rs375186872 Chr.10: 95042899-95042899 on Build GRCh38
rs772656000 Chr.10: 95042893-95042893 on Build GRCh38
rs945187704 Chr.10: 95042934-95042934 on Build GRCh38
rs760378028 Chr.10: 95042810-95042810 on Build GRCh38
rs991543575 Chr.10: 95042820-95042820 on Build GRCh38
rs774260681 Chr.10: 95042723-95042723 on Build GRCh38
rs200115418 Chr.10: 95042871-95042871 on Build GRCh38
rs77147096 Chr.10: 95042946-95042946 on Build GRCh38
rs759186099 Chr.10: 95042903-95042903 on Build GRCh38
rs79270715 Chr.10: 95042819-95042819 on Build GRCh38
rs765139666 Chr.10: 95042891-95042891 on Build GRCh38
rs760330613 Chr.10: 95042898-95042898 on Build GRCh38
rs371605006 Chr.10: 95042886-95042886 on Build GRCh38
rs779601447 Chr.10: 95042858-95042858 on Build GRCh38
rs569528471 Chr.10: 95042753-95042753 on Build GRCh38
rs943138692 Chr.10: 95042927-95042927 on Build GRCh38
rs542760737 Chr.10: 95042885-95042885 on Build GRCh38
rs371715037 Chr.10: 95042867-95042867 on Build GRCh38
rs186219558 Chr.10: 95042733-95042733 on Build GRCh38
rs375273765 Chr.10: 95042869-95042869 on Build GRCh38
rs760811547 Chr.10: 95042875-95042875 on Build GRCh38
rs1000748565 Chr.10: 95042949-95042949 on Build GRCh38
rs764839435 Chr.10: 95042925-95042925 on Build GRCh38
rs761921124 Chr.10: 95042887-95042887 on Build GRCh38
rs375477204 Chr.10: 95042839-95042839 on Build GRCh38
rs772685066 Chr.10: 95042804-95042804 on Build GRCh38
rs1030435423 Chr.10: 95042740-95042740 on Build GRCh38
rs771598475 Chr.10: 95042862-95042867 on Build GRCh38
rs147133669 Chr.10: 95042943-95042943 on Build GRCh38

View Full Product Details