Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 25170422-25170649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER2
Entrez Gene ID
219287
Gene Name
APC membrane recruitment protein 2
Gene Aliases
FAM123A
UniGene
Hs.528335
Gene Chromosome Location
Chr.13: 25161679-25172456 on Build GRCh38
Transcripts
NM_152704.3 NM_199138.2 XM_005266279.3 XM_005266280.3 XM_017020415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1300097 c.814G>C p.E272Q Chr.13: 25170449-25170449 on Build GRCh38
7273705 c.1150A>T p.I384F Chr.13: 25170470-25170470 on Build GRCh38
7265446 c.1166A>C p.E389A Chr.13: 25170454-25170454 on Build GRCh38
6520930 c.1022G>T p.R341L Chr.13: 25170598-25170598 on Build GRCh38
4046721 c.829T>C p.C277R Chr.13: 25170434-25170434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013140561 Chr.13: 25170571-25170571 on Build GRCh38
rs745614355 Chr.13: 25170474-25170474 on Build GRCh38
rs778017679 Chr.13: 25170549-25170549 on Build GRCh38
rs748931960 Chr.13: 25170499-25170499 on Build GRCh38
rs749700120 Chr.13: 25170595-25170595 on Build GRCh38
rs776925046 Chr.13: 25170627-25170627 on Build GRCh38
rs771589115 Chr.13: 25170610-25170610 on Build GRCh38
rs764116167 Chr.13: 25170566-25170566 on Build GRCh38
rs946379280 Chr.13: 25170550-25170550 on Build GRCh38
rs747570250 Chr.13: 25170606-25170606 on Build GRCh38
rs199586338 Chr.13: 25170467-25170467 on Build GRCh38
rs759919827 Chr.13: 25170628-25170628 on Build GRCh38
rs778649468 Chr.13: 25170594-25170594 on Build GRCh38
rs756174964 Chr.13: 25170573-25170573 on Build GRCh38
rs750360539 Chr.13: 25170567-25170567 on Build GRCh38
rs1035181364 Chr.13: 25170592-25170592 on Build GRCh38
rs911749540 Chr.13: 25170472-25170472 on Build GRCh38
rs749098220 Chr.13: 25170588-25170588 on Build GRCh38
rs765512835 Chr.13: 25170555-25170555 on Build GRCh38
rs754618693 Chr.13: 25170593-25170593 on Build GRCh38
rs773799706 Chr.13: 25170536-25170536 on Build GRCh38
rs1042127438 Chr.13: 25170560-25170560 on Build GRCh38
rs771434632 Chr.13: 25170626-25170626 on Build GRCh38
rs370004072 Chr.13: 25170518-25170518 on Build GRCh38
rs377253214 Chr.13: 25170446-25170446 on Build GRCh38
rs200938266 Chr.13: 25170547-25170547 on Build GRCh38
rs772656513 Chr.13: 25170611-25170611 on Build GRCh38
rs796490688 Chr.13: 25170477-25170477 on Build GRCh38
rs768486921 Chr.13: 25170506-25170506 on Build GRCh38
rs201615060 Chr.13: 25170488-25170488 on Build GRCh38
rs752673606 Chr.13: 25170561-25170561 on Build GRCh38
rs747512089 Chr.13: 25170613-25170613 on Build GRCh38
rs375802560 Chr.13: 25170562-25170562 on Build GRCh38
rs772409592 Chr.13: 25170520-25170520 on Build GRCh38
rs779988402 Chr.13: 25170587-25170587 on Build GRCh38

View Full Product Details