Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75104862-75105101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD31
Entrez Gene ID
256006
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
UniGene
Hs.482533
Gene Chromosome Location
Chr.5: 75068297-75236897 on Build GRCh38
Transcripts
NM_001164443.1 XM_011543298.2 XM_011543299.1 XM_011543300.2 XM_011543301.2 XM_011543302.2 XM_017009317.1 XM_017009318.1 XM_017009319.1 XM_017009320.1 XM_017009321.1 XM_017009322.1

SNP Detail

SNP ID Chromosome Location
rs754709554 Chr.5: 75105042-75105042 on Build GRCh38
rs532616902 Chr.5: 75104906-75104906 on Build GRCh38
rs1003788379 Chr.5: 75104980-75104980 on Build GRCh38
rs543209682 Chr.5: 75105017-75105017 on Build GRCh38
rs898021118 Chr.5: 75105018-75105018 on Build GRCh38
rs866769810 Chr.5: 75105064-75105064 on Build GRCh38
rs756097632 Chr.5: 75105050-75105050 on Build GRCh38
rs767348382 Chr.5: 75104979-75104979 on Build GRCh38
rs765603500 Chr.5: 75104930-75104930 on Build GRCh38
rs986200921 Chr.5: 75105043-75105043 on Build GRCh38
rs181179046 Chr.5: 75104958-75104958 on Build GRCh38
rs994052265 Chr.5: 75105019-75105019 on Build GRCh38
rs760576408 Chr.5: 75104940-75104940 on Build GRCh38
rs750128438 Chr.5: 75104992-75104992 on Build GRCh38

View Full Product Details