Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110212066-110212323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC40
Entrez Gene ID
51362
Gene Name
cell division cycle 40
Gene Aliases
EHB3, PRP17, PRPF17
UniGene
Hs.428147
Gene Chromosome Location
Chr.6: 110180421-110232220 on Build GRCh38
Transcripts
NM_015891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1439725 c.839A>G p.Q280R Chr.6: 110212244-110212244 on Build GRCh38
4760074 c.832A>G p.K278E Chr.6: 110212237-110212237 on Build GRCh38
4760072 c.830C>G p.P277R Chr.6: 110212235-110212235 on Build GRCh38
5468462 c.862A>C p.T288P Chr.6: 110212267-110212267 on Build GRCh38
5881035 c.755G>T p.R252M Chr.6: 110212160-110212160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747870092 Chr.6: 110212268-110212268 on Build GRCh38
rs200012582 Chr.6: 110212103-110212103 on Build GRCh38
rs890438403 Chr.6: 110212183-110212183 on Build GRCh38
rs757888735 Chr.6: 110212252-110212252 on Build GRCh38
rs774620104 Chr.6: 110212166-110212166 on Build GRCh38
rs769922530 Chr.6: 110212278-110212278 on Build GRCh38
rs760746039 Chr.6: 110212202-110212202 on Build GRCh38
rs376762186 Chr.6: 110212158-110212158 on Build GRCh38
rs1051986264 Chr.6: 110212161-110212161 on Build GRCh38
rs144149633 Chr.6: 110212194-110212194 on Build GRCh38
rs781060864 Chr.6: 110212262-110212262 on Build GRCh38
rs759687430 Chr.6: 110212173-110212173 on Build GRCh38
rs1039110880 Chr.6: 110212287-110212287 on Build GRCh38
rs1028720077 Chr.6: 110212184-110212184 on Build GRCh38
rs374768334 Chr.6: 110212210-110212210 on Build GRCh38
rs778419391 Chr.6: 110212112-110212112 on Build GRCh38
rs768509564 Chr.6: 110212155-110212155 on Build GRCh38
rs757664959 Chr.6: 110212104-110212104 on Build GRCh38
rs745361098 Chr.6: 110212119-110212119 on Build GRCh38
rs373546052 Chr.6: 110212165-110212165 on Build GRCh38
rs878893916 Chr.6: 110212261-110212261 on Build GRCh38
rs924812651 Chr.6: 110212118-110212118 on Build GRCh38
rs754103178 Chr.6: 110212208-110212208 on Build GRCh38
rs140630036 Chr.6: 110212143-110212143 on Build GRCh38
rs752985644 Chr.6: 110212193-110212193 on Build GRCh38
rs771751370 Chr.6: 110212140-110212140 on Build GRCh38
rs779362671 Chr.6: 110212230-110212230 on Build GRCh38
rs746677855 Chr.6: 110212254-110212254 on Build GRCh38
rs767912290 Chr.6: 110212192-110212192 on Build GRCh38
rs147439944 Chr.6: 110212207-110212207 on Build GRCh38
rs773271331 Chr.6: 110212284-110212284 on Build GRCh38
rs746838076 Chr.6: 110212144-110212144 on Build GRCh38
rs750001687 Chr.6: 110212241-110212241 on Build GRCh38
rs776219148 Chr.6: 110212157-110212157 on Build GRCh38
rs369306851 Chr.6: 110212095-110212095 on Build GRCh38

View Full Product Details