Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147881753-147882022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP10
Entrez Gene ID
79658
Gene Name
Rho GTPase activating protein 10
Gene Aliases
GRAF2, PS-GAP, PSGAP
UniGene
Hs.368631
Gene Chromosome Location
Chr.4: 147732063-148072781 on Build GRCh38
Transcripts
NM_024605.3 XM_005263215.3 XM_017008602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919802 c.946G>T p.G316* Chr.4: 147881844-147881844 on Build GRCh38
3767722 c.945C>G p.D315E Chr.4: 147881843-147881843 on Build GRCh38
3127226 c.946G>A p.G316R Chr.4: 147881844-147881844 on Build GRCh38
6603013 c.1034G>A p.R345Q Chr.4: 147881932-147881932 on Build GRCh38
6535461 c.1020A>G p.I340M Chr.4: 147881918-147881918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761358510 Chr.4: 147881959-147881960 on Build GRCh38
rs898026652 Chr.4: 147881778-147881778 on Build GRCh38
rs759250334 Chr.4: 147881932-147881932 on Build GRCh38
rs753305749 Chr.4: 147881840-147881840 on Build GRCh38
rs573781469 Chr.4: 147881975-147881975 on Build GRCh38
rs748823657 Chr.4: 147881910-147881910 on Build GRCh38
rs989980468 Chr.4: 147881917-147881917 on Build GRCh38
rs769400769 Chr.4: 147881799-147881799 on Build GRCh38
rs1014704004 Chr.4: 147881881-147881881 on Build GRCh38
rs761882313 Chr.4: 147881959-147881959 on Build GRCh38
rs955894117 Chr.4: 147881916-147881916 on Build GRCh38
rs768379995 Chr.4: 147881789-147881789 on Build GRCh38
rs748027857 Chr.4: 147881798-147881798 on Build GRCh38
rs769655337 Chr.4: 147881893-147881893 on Build GRCh38
rs1030002467 Chr.4: 147881821-147881821 on Build GRCh38
rs900118362 Chr.4: 147881783-147881783 on Build GRCh38
rs374199958 Chr.4: 147881797-147881797 on Build GRCh38
rs1027899515 Chr.4: 147881816-147881816 on Build GRCh38
rs115776553 Chr.4: 147881956-147881956 on Build GRCh38
rs377621378 Chr.4: 147881822-147881822 on Build GRCh38
rs201421446 Chr.4: 147881875-147881875 on Build GRCh38
rs759986173 Chr.4: 147881830-147881830 on Build GRCh38
rs770733120 Chr.4: 147881815-147881815 on Build GRCh38
rs549345257 Chr.4: 147881839-147881839 on Build GRCh38
rs551383996 Chr.4: 147881953-147881953 on Build GRCh38
rs61758694 Chr.4: 147881876-147881876 on Build GRCh38
rs772646380 Chr.4: 147881974-147881975 on Build GRCh38
rs776437321 Chr.4: 147881957-147881958 on Build GRCh38
rs770219638 Chr.4: 147881843-147881843 on Build GRCh38
rs777720441 Chr.4: 147881898-147881898 on Build GRCh38
rs748161616 Chr.4: 147881892-147881892 on Build GRCh38
rs138388284 Chr.4: 147881844-147881844 on Build GRCh38
rs772614370 Chr.4: 147881939-147881939 on Build GRCh38
rs992681079 Chr.4: 147881988-147881988 on Build GRCh38
rs770500236 Chr.4: 147881919-147881919 on Build GRCh38
rs376444955 Chr.4: 147881970-147881970 on Build GRCh38
rs753893705 Chr.4: 147881854-147881854 on Build GRCh38
rs993617359 Chr.4: 147881794-147881794 on Build GRCh38
rs17024037 Chr.4: 147881950-147881950 on Build GRCh38
rs768358224 Chr.4: 147881836-147881837 on Build GRCh38
rs762489163 Chr.4: 147881813-147881813 on Build GRCh38
rs754830371 Chr.4: 147881889-147881889 on Build GRCh38
rs779961144 Chr.4: 147881931-147881931 on Build GRCh38
rs200501384 Chr.4: 147881977-147881977 on Build GRCh38
rs373578495 Chr.4: 147881967-147881967 on Build GRCh38
rs780831683 Chr.4: 147881891-147881891 on Build GRCh38
rs746306875 Chr.4: 147881841-147881841 on Build GRCh38
rs757275728 Chr.4: 147881873-147881873 on Build GRCh38
rs932570612 Chr.4: 147881933-147881933 on Build GRCh38
rs1013133448 Chr.4: 147881826-147881826 on Build GRCh38

View Full Product Details