Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151048214-151048441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf56
Entrez Gene ID
54964
Gene Name
chromosome 1 open reading frame 56
Gene Aliases
MENT
UniGene
Hs.549171
Gene Chromosome Location
Chr.1: 151047783-151051420 on Build GRCh38
Transcripts
NM_017860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
352814 c.455C>T p.S152L Chr.1: 151048302-151048302 on Build GRCh38
6272730 c.535G>T p.G179W Chr.1: 151048382-151048382 on Build GRCh38
895687 c.386C>T p.P129L Chr.1: 151048233-151048233 on Build GRCh38
1240927 c.542delC p.P182fs*22 Chr.1: 151048389-151048389 on Build GRCh38
3473573 c.519C>G p.S173R Chr.1: 151048366-151048366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986822059 Chr.1: 151048388-151048388 on Build GRCh38
rs923365563 Chr.1: 151048377-151048377 on Build GRCh38
rs764050067 Chr.1: 151048381-151048381 on Build GRCh38
rs757691338 Chr.1: 151048417-151048417 on Build GRCh38
rs752910122 Chr.1: 151048374-151048374 on Build GRCh38
rs779396881 Chr.1: 151048422-151048422 on Build GRCh38
rs761344402 Chr.1: 151048317-151048317 on Build GRCh38
rs756197124 Chr.1: 151048313-151048313 on Build GRCh38
rs766053184 Chr.1: 151048291-151048291 on Build GRCh38
rs761322124 Chr.1: 151048332-151048332 on Build GRCh38
rs200269793 Chr.1: 151048354-151048354 on Build GRCh38
rs972000542 Chr.1: 151048324-151048324 on Build GRCh38
rs368980042 Chr.1: 151048311-151048311 on Build GRCh38
rs35890495 Chr.1: 151048242-151048243 on Build GRCh38
rs771684321 Chr.1: 151048397-151048397 on Build GRCh38
rs369604893 Chr.1: 151048328-151048328 on Build GRCh38
rs986214244 Chr.1: 151048389-151048389 on Build GRCh38
rs746719165 Chr.1: 151048322-151048322 on Build GRCh38
rs776103203 Chr.1: 151048253-151048253 on Build GRCh38
rs1046586150 Chr.1: 151048240-151048240 on Build GRCh38
rs761804232 Chr.1: 151048254-151048254 on Build GRCh38
rs150905834 Chr.1: 151048341-151048341 on Build GRCh38
rs191147545 Chr.1: 151048236-151048236 on Build GRCh38
rs779123317 Chr.1: 151048319-151048319 on Build GRCh38
rs149675198 Chr.1: 151048380-151048380 on Build GRCh38
rs759906547 Chr.1: 151048371-151048371 on Build GRCh38
rs781481167 Chr.1: 151048307-151048307 on Build GRCh38
rs768202321 Chr.1: 151048248-151048248 on Build GRCh38
rs772795385 Chr.1: 151048413-151048413 on Build GRCh38
rs750267438 Chr.1: 151048270-151048270 on Build GRCh38
rs776194426 Chr.1: 151048331-151048331 on Build GRCh38
rs34253426 Chr.1: 151048321-151048322 on Build GRCh38
rs112832558 Chr.1: 151048330-151048330 on Build GRCh38
rs765220707 Chr.1: 151048265-151048265 on Build GRCh38
rs899458649 Chr.1: 151048287-151048287 on Build GRCh38
rs774276719 Chr.1: 151048358-151048358 on Build GRCh38
rs767857236 Chr.1: 151048373-151048373 on Build GRCh38
rs778359132 Chr.1: 151048314-151048314 on Build GRCh38
rs760287849 Chr.1: 151048243-151048243 on Build GRCh38
rs771364471 Chr.1: 151048317-151048317 on Build GRCh38
rs769140079 Chr.1: 151048338-151048338 on Build GRCh38
rs762833017 Chr.1: 151048272-151048272 on Build GRCh38
rs587603472 Chr.1: 151048274-151048277 on Build GRCh38
rs755232808 Chr.1: 151048305-151048305 on Build GRCh38

View Full Product Details