Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 165829815-165830076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCHE
Entrez Gene ID
590
Gene Name
butyrylcholinesterase
Gene Aliases
CHE1, CHE2, E1
UniGene
Hs.420483
Gene Chromosome Location
Chr.3: 165772904-165837467 on Build GRCh38
Transcripts
XM_017006967.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742665 c.1207G>T p.D403Y Chr.3: 165829827-165829827 on Build GRCh38
1168986 c.1130A>T p.E377V Chr.3: 165829904-165829904 on Build GRCh38
4606932 c.1020T>G p.F340L Chr.3: 165830014-165830014 on Build GRCh38
7245006 c.963C>A p.T321T Chr.3: 165830071-165830071 on Build GRCh38
149507 c.1199A>G p.H400R Chr.3: 165829835-165829835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996521009 Chr.3: 165830038-165830038 on Build GRCh38
rs766000843 Chr.3: 165830047-165830047 on Build GRCh38
rs763000328 Chr.3: 165829893-165829893 on Build GRCh38
rs763806377 Chr.3: 165829925-165829925 on Build GRCh38
rs115129687 Chr.3: 165829857-165829857 on Build GRCh38
rs747438953 Chr.3: 165829856-165829856 on Build GRCh38
rs765249814 Chr.3: 165829943-165829943 on Build GRCh38
rs537434945 Chr.3: 165830007-165830007 on Build GRCh38
rs753284945 Chr.3: 165829925-165829927 on Build GRCh38
rs767469658 Chr.3: 165829875-165829876 on Build GRCh38
rs776350992 Chr.3: 165830021-165830021 on Build GRCh38
rs764498866 Chr.3: 165829999-165830007 on Build GRCh38
rs1057516450 Chr.3: 165829960-165829961 on Build GRCh38
rs750000733 Chr.3: 165829944-165829944 on Build GRCh38
rs372839550 Chr.3: 165829897-165829897 on Build GRCh38
rs373114728 Chr.3: 165829892-165829892 on Build GRCh38
rs1022014694 Chr.3: 165829970-165829970 on Build GRCh38
rs748533851 Chr.3: 165829966-165829966 on Build GRCh38
rs104893684 Chr.3: 165830030-165830030 on Build GRCh38
rs150912772 Chr.3: 165830042-165830042 on Build GRCh38
rs774590309 Chr.3: 165829900-165829900 on Build GRCh38
rs765307050 Chr.3: 165830037-165830037 on Build GRCh38
rs772337347 Chr.3: 165830013-165830013 on Build GRCh38
rs748608343 Chr.3: 165829888-165829888 on Build GRCh38
rs1016349008 Chr.3: 165829922-165829922 on Build GRCh38
rs754214624 Chr.3: 165830006-165830007 on Build GRCh38
rs1057517265 Chr.3: 165830019-165830019 on Build GRCh38
rs376091945 Chr.3: 165829963-165829963 on Build GRCh38
rs781476667 Chr.3: 165829965-165829965 on Build GRCh38
rs749752833 Chr.3: 165829995-165829995 on Build GRCh38
rs899539416 Chr.3: 165830005-165830005 on Build GRCh38
rs759679732 Chr.3: 165829860-165829863 on Build GRCh38
rs139312740 Chr.3: 165829939-165829939 on Build GRCh38
rs1057517221 Chr.3: 165829851-165829851 on Build GRCh38
rs778225256 Chr.3: 165829981-165829981 on Build GRCh38
rs759686297 Chr.3: 165830012-165830012 on Build GRCh38
rs146936556 Chr.3: 165829884-165829884 on Build GRCh38
rs143300460 Chr.3: 165829957-165829957 on Build GRCh38
rs201120931 Chr.3: 165829956-165829956 on Build GRCh38
rs753598758 Chr.3: 165829931-165829931 on Build GRCh38
rs770337031 Chr.3: 165829967-165829967 on Build GRCh38
rs753657650 Chr.3: 165829974-165829974 on Build GRCh38
rs534912670 Chr.3: 165829905-165829905 on Build GRCh38
rs757948667 Chr.3: 165829947-165829947 on Build GRCh38
rs761080105 Chr.3: 165829932-165829932 on Build GRCh38
rs771017682 Chr.3: 165829996-165829996 on Build GRCh38
rs557112656 Chr.3: 165830014-165830014 on Build GRCh38
rs121918557 Chr.3: 165829962-165829962 on Build GRCh38

View Full Product Details