Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27547824-27548077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHDC1
Entrez Gene ID
27245
Gene Name
AT-hook DNA binding motif containing 1
Gene Aliases
MRD25
UniGene
Hs.469280
Gene Chromosome Location
Chr.1: 27534245-27604178 on Build GRCh38
Transcripts
NM_001029882.3 XM_005245848.3 XM_005245849.3 XM_005245850.3 XM_005245851.3 XM_005245852.3 XM_011541255.1 XM_011541256.2 XM_011541257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6314107 c.4098C>A p.D1366E Chr.1: 27548018-27548018 on Build GRCh38
4492924 c.4064C>T p.S1355F Chr.1: 27548052-27548052 on Build GRCh38
4493667 c.4199C>T p.P1400L Chr.1: 27547917-27547917 on Build GRCh38
5215424 c.4176_4177insC p.G1393fs*88 Chr.1: 27547939-27547940 on Build GRCh38
6266770 c.4151C>A p.A1384D Chr.1: 27547965-27547965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547570973 Chr.1: 27547862-27547862 on Build GRCh38
rs754747978 Chr.1: 27547965-27547965 on Build GRCh38
rs757367512 Chr.1: 27548058-27548058 on Build GRCh38
rs148277570 Chr.1: 27547938-27547938 on Build GRCh38
rs748755041 Chr.1: 27547999-27547999 on Build GRCh38
rs758382362 Chr.1: 27547959-27547959 on Build GRCh38
rs962169806 Chr.1: 27548050-27548050 on Build GRCh38
rs982121315 Chr.1: 27548027-27548027 on Build GRCh38
rs745791429 Chr.1: 27547890-27547890 on Build GRCh38
rs763438140 Chr.1: 27547933-27547933 on Build GRCh38
rs142913680 Chr.1: 27547853-27547853 on Build GRCh38
rs772776553 Chr.1: 27547925-27547925 on Build GRCh38
rs780103526 Chr.1: 27547961-27547961 on Build GRCh38
rs779976259 Chr.1: 27547896-27547896 on Build GRCh38
rs910475305 Chr.1: 27547964-27547964 on Build GRCh38
rs1012505411 Chr.1: 27547997-27547997 on Build GRCh38
rs919736278 Chr.1: 27547922-27547922 on Build GRCh38
rs754016013 Chr.1: 27547957-27547957 on Build GRCh38
rs750886804 Chr.1: 27547855-27547855 on Build GRCh38
rs756355656 Chr.1: 27547953-27547953 on Build GRCh38
rs952050939 Chr.1: 27547942-27547942 on Build GRCh38
rs747832311 Chr.1: 27547971-27547971 on Build GRCh38
rs753474234 Chr.1: 27548020-27548020 on Build GRCh38
rs752894663 Chr.1: 27547952-27547952 on Build GRCh38
rs375964883 Chr.1: 27547850-27547850 on Build GRCh38
rs775771211 Chr.1: 27548025-27548025 on Build GRCh38
rs764216089 Chr.1: 27547955-27547955 on Build GRCh38
rs777716141 Chr.1: 27547879-27547879 on Build GRCh38
rs201742929 Chr.1: 27547871-27547871 on Build GRCh38
rs764454493 Chr.1: 27548042-27548042 on Build GRCh38
rs984798332 Chr.1: 27547956-27547956 on Build GRCh38
rs758464071 Chr.1: 27547895-27547895 on Build GRCh38
rs766726003 Chr.1: 27547860-27547860 on Build GRCh38
rs774782166 Chr.1: 27548015-27548015 on Build GRCh38
rs770447679 Chr.1: 27548011-27548011 on Build GRCh38
rs141082682 Chr.1: 27547872-27547872 on Build GRCh38
rs371911768 Chr.1: 27547939-27547939 on Build GRCh38
rs147174735 Chr.1: 27548057-27548057 on Build GRCh38
rs199996166 Chr.1: 27547981-27547981 on Build GRCh38
rs1044392390 Chr.1: 27547884-27547884 on Build GRCh38
rs756159146 Chr.1: 27547878-27547878 on Build GRCh38
rs763456777 Chr.1: 27547858-27547858 on Build GRCh38
rs527518359 Chr.1: 27547847-27547847 on Build GRCh38
rs775977549 Chr.1: 27547849-27547849 on Build GRCh38
rs145735811 Chr.1: 27547913-27547913 on Build GRCh38
rs765921462 Chr.1: 27547996-27547996 on Build GRCh38
rs769391540 Chr.1: 27547976-27547976 on Build GRCh38
rs149518384 Chr.1: 27547916-27547916 on Build GRCh38
rs767916808 Chr.1: 27548021-27548021 on Build GRCh38
rs146774132 Chr.1: 27547898-27547898 on Build GRCh38
rs760926906 Chr.1: 27548028-27548028 on Build GRCh38
rs143406235 Chr.1: 27548030-27548030 on Build GRCh38
rs893943868 Chr.1: 27547982-27547982 on Build GRCh38
rs767786022 Chr.1: 27547940-27547940 on Build GRCh38
rs886044186 Chr.1: 27548043-27548043 on Build GRCh38
rs769282629 Chr.1: 27547919-27547919 on Build GRCh38
rs374097223 Chr.1: 27547937-27547937 on Build GRCh38
rs147352724 Chr.1: 27547970-27547970 on Build GRCh38
rs745330577 Chr.1: 27547907-27547907 on Build GRCh38

View Full Product Details