Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222921357-222921616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL3
Entrez Gene ID
2181
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
ACS3, FACL3, PRO2194
UniGene
Hs.655772
Gene Chromosome Location
Chr.2: 222860984-222944624 on Build GRCh38
Transcripts
NM_004457.3 NM_203372.1 XM_017003584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5908053 c.941G>A p.R314K Chr.2: 222921415-222921415 on Build GRCh38
1017053 c.921T>C p.G307G Chr.2: 222921395-222921395 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs961576218 Chr.2: 222921577-222921577 on Build GRCh38
rs759815086 Chr.2: 222921440-222921440 on Build GRCh38
rs1028549420 Chr.2: 222921498-222921498 on Build GRCh38
rs186465496 Chr.2: 222921470-222921470 on Build GRCh38
rs748143189 Chr.2: 222921410-222921410 on Build GRCh38
rs766913664 Chr.2: 222921479-222921479 on Build GRCh38
rs775855621 Chr.2: 222921451-222921451 on Build GRCh38
rs891385505 Chr.2: 222921525-222921525 on Build GRCh38
rs770245610 Chr.2: 222921558-222921559 on Build GRCh38
rs1017500952 Chr.2: 222921538-222921538 on Build GRCh38
rs749993660 Chr.2: 222921480-222921480 on Build GRCh38
rs1031402645 Chr.2: 222921404-222921404 on Build GRCh38
rs80245918 Chr.2: 222921545-222921545 on Build GRCh38
rs1043822104 Chr.2: 222921389-222921389 on Build GRCh38
rs763348450 Chr.2: 222921463-222921463 on Build GRCh38
rs776815801 Chr.2: 222921427-222921427 on Build GRCh38
rs769065377 Chr.2: 222921479-222921480 on Build GRCh38
rs762421760 Chr.2: 222921472-222921472 on Build GRCh38
rs531543148 Chr.2: 222921519-222921519 on Build GRCh38
rs770879420 Chr.2: 222921412-222921412 on Build GRCh38
rs764538983 Chr.2: 222921466-222921466 on Build GRCh38
rs138183640 Chr.2: 222921550-222921550 on Build GRCh38
rs547485872 Chr.2: 222921529-222921529 on Build GRCh38
rs770203903 Chr.2: 222921441-222921441 on Build GRCh38
rs781160116 Chr.2: 222921527-222921527 on Build GRCh38

View Full Product Details