Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37621022-37621255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX52
Entrez Gene ID
11056
Gene Name
DEAD-box helicase 52
Gene Aliases
HUSSY19, ROK1
UniGene
Hs.590937
Gene Chromosome Location
Chr.17: 37609739-37643464 on Build GRCh38
Transcripts
NM_001291476.1 NM_007010.4 XM_011524232.2 XM_011524233.2

SNP Detail

SNP ID Chromosome Location
rs867130838 Chr.17: 37621144-37621144 on Build GRCh38
rs747593652 Chr.17: 37621108-37621109 on Build GRCh38
rs368118442 Chr.17: 37621179-37621179 on Build GRCh38
rs747612415 Chr.17: 37621169-37621169 on Build GRCh38
rs779958448 Chr.17: 37621156-37621156 on Build GRCh38
rs912684745 Chr.17: 37621073-37621073 on Build GRCh38
rs373687391 Chr.17: 37621086-37621086 on Build GRCh38
rs146778713 Chr.17: 37621215-37621215 on Build GRCh38
rs773644959 Chr.17: 37621202-37621202 on Build GRCh38
rs563719400 Chr.17: 37621065-37621065 on Build GRCh38
rs778568642 Chr.17: 37621168-37621168 on Build GRCh38
rs771183934 Chr.17: 37621218-37621218 on Build GRCh38
rs752860803 Chr.17: 37621087-37621087 on Build GRCh38
rs914540745 Chr.17: 37621132-37621132 on Build GRCh38
rs772348428 Chr.17: 37621078-37621078 on Build GRCh38
rs180970089 Chr.17: 37621151-37621151 on Build GRCh38
rs758581649 Chr.17: 37621090-37621090 on Build GRCh38
rs746058731 Chr.17: 37621053-37621053 on Build GRCh38
rs988811707 Chr.17: 37621102-37621102 on Build GRCh38
rs986972824 Chr.17: 37621062-37621062 on Build GRCh38
rs938549884 Chr.17: 37621197-37621197 on Build GRCh38
rs748871938 Chr.17: 37621157-37621157 on Build GRCh38
rs529308402 Chr.17: 37621069-37621069 on Build GRCh38
rs781226062 Chr.17: 37621107-37621107 on Build GRCh38
rs947336309 Chr.17: 37621137-37621137 on Build GRCh38
rs771587883 Chr.17: 37621200-37621200 on Build GRCh38
rs1010271839 Chr.17: 37621106-37621106 on Build GRCh38
rs749398921 Chr.17: 37621070-37621070 on Build GRCh38
rs768395472 Chr.17: 37621158-37621158 on Build GRCh38
rs769023340 Chr.17: 37621109-37621109 on Build GRCh38
rs956156942 Chr.17: 37621074-37621074 on Build GRCh38
rs934728362 Chr.17: 37621082-37621082 on Build GRCh38
rs750397580 Chr.17: 37621117-37621117 on Build GRCh38
rs61756268 Chr.17: 37621227-37621227 on Build GRCh38

View Full Product Details