Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107931956-107932183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2 NR_125929.1

Gene Symbol
LOC101929595
Entrez Gene ID
101929595
Gene Name
uncharacterized LOC101929595
Gene Aliases
-
UniGene
Hs.678014
Gene Chromosome Location
Chr.4: 107863479-107932119 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2 NR_125929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619819 c.400C>T p.H134Y Chr.4: 107932043-107932043 on Build GRCh38
6497794 c.409C>T p.R137C Chr.4: 107932052-107932052 on Build GRCh38
6289641 c.351C>T p.I117I Chr.4: 107931994-107931994 on Build GRCh38
6218857 c.395A>T p.D132V Chr.4: 107932038-107932038 on Build GRCh38
6512442 c.470C>A p.S157Y Chr.4: 107932113-107932113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021034246 Chr.4: 107931985-107931985 on Build GRCh38
rs760309229 Chr.4: 107932084-107932084 on Build GRCh38
rs763896315 Chr.4: 107932001-107932001 on Build GRCh38
rs140010604 Chr.4: 107932068-107932068 on Build GRCh38
rs893894789 Chr.4: 107932079-107932079 on Build GRCh38
rs747178895 Chr.4: 107932043-107932043 on Build GRCh38
rs750275646 Chr.4: 107932024-107932024 on Build GRCh38
rs557330754 Chr.4: 107932162-107932162 on Build GRCh38
rs761575210 Chr.4: 107932095-107932095 on Build GRCh38
rs774860849 Chr.4: 107932082-107932082 on Build GRCh38
rs1023463496 Chr.4: 107932097-107932097 on Build GRCh38
rs766889023 Chr.4: 107931986-107931986 on Build GRCh38
rs773648558 Chr.4: 107932060-107932060 on Build GRCh38
rs781445230 Chr.4: 107932046-107932046 on Build GRCh38
rs771474750 Chr.4: 107932069-107932069 on Build GRCh38
rs768640920 Chr.4: 107932113-107932113 on Build GRCh38
rs983198585 Chr.4: 107932125-107932125 on Build GRCh38
rs971295974 Chr.4: 107932014-107932014 on Build GRCh38
rs762792329 Chr.4: 107932109-107932109 on Build GRCh38
rs368014569 Chr.4: 107932039-107932039 on Build GRCh38
rs781284978 Chr.4: 107932151-107932151 on Build GRCh38
rs755182820 Chr.4: 107932044-107932044 on Build GRCh38
rs756343727 Chr.4: 107932163-107932163 on Build GRCh38
rs942947151 Chr.4: 107932019-107932019 on Build GRCh38
rs770108230 Chr.4: 107932059-107932059 on Build GRCh38
rs763547405 Chr.4: 107931979-107931979 on Build GRCh38
rs776382381 Chr.4: 107932093-107932093 on Build GRCh38
rs760339720 Chr.4: 107931994-107931994 on Build GRCh38
rs748485159 Chr.4: 107932055-107932055 on Build GRCh38
rs1048541745 Chr.4: 107932013-107932013 on Build GRCh38
rs750234190 Chr.4: 107932103-107932103 on Build GRCh38
rs776652155 Chr.4: 107932138-107932139 on Build GRCh38
rs375489915 Chr.4: 107932015-107932015 on Build GRCh38
rs1003478972 Chr.4: 107932153-107932153 on Build GRCh38
rs748099773 Chr.4: 107932139-107932140 on Build GRCh38
rs147612190 Chr.4: 107932139-107932139 on Build GRCh38
rs577594501 Chr.4: 107932030-107932030 on Build GRCh38
rs752773819 Chr.4: 107932063-107932063 on Build GRCh38
rs757157523 Chr.4: 107932004-107932004 on Build GRCh38
rs1034328850 Chr.4: 107932035-107932035 on Build GRCh38
rs768390894 Chr.4: 107932090-107932090 on Build GRCh38
rs959668671 Chr.4: 107932036-107932036 on Build GRCh38
rs920309099 Chr.4: 107931991-107931991 on Build GRCh38
rs766399892 Chr.4: 107932123-107932123 on Build GRCh38
rs753662731 Chr.4: 107932003-107932003 on Build GRCh38
rs764878369 Chr.4: 107932096-107932096 on Build GRCh38
rs747481440 Chr.4: 107932125-107932127 on Build GRCh38

View Full Product Details