Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27598134-27598377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLU
Entrez Gene ID
1191
Gene Name
clusterin
Gene Aliases
AAG4, APO-J, APOJ, CLI, CLU1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2
UniGene
Hs.436657
Gene Chromosome Location
Chr.8: 27596917-27616717 on Build GRCh38
Transcripts
NM_001831.3 NR_038335.1 NR_045494.1 XM_006716284.2

SNP Detail

SNP ID Chromosome Location
rs373382856 Chr.8: 27598153-27598153 on Build GRCh38
rs752908775 Chr.8: 27598194-27598194 on Build GRCh38
rs975611826 Chr.8: 27598354-27598354 on Build GRCh38
rs200244739 Chr.8: 27598232-27598232 on Build GRCh38
rs954638755 Chr.8: 27598229-27598229 on Build GRCh38
rs759248863 Chr.8: 27598242-27598242 on Build GRCh38
rs534510358 Chr.8: 27598350-27598350 on Build GRCh38
rs746510318 Chr.8: 27598168-27598168 on Build GRCh38
rs772897252 Chr.8: 27598288-27598288 on Build GRCh38
rs1018541329 Chr.8: 27598175-27598175 on Build GRCh38
rs927250516 Chr.8: 27598344-27598344 on Build GRCh38
rs780113491 Chr.8: 27598233-27598233 on Build GRCh38
rs748619722 Chr.8: 27598189-27598191 on Build GRCh38
rs372604797 Chr.8: 27598278-27598278 on Build GRCh38
rs779303630 Chr.8: 27598189-27598189 on Build GRCh38
rs1042830481 Chr.8: 27598325-27598325 on Build GRCh38
rs772456164 Chr.8: 27598257-27598258 on Build GRCh38
rs75016658 Chr.8: 27598324-27598324 on Build GRCh38
rs370655263 Chr.8: 27598192-27598192 on Build GRCh38
rs13127 Chr.8: 27598206-27598206 on Build GRCh38
rs1030956980 Chr.8: 27598205-27598206 on Build GRCh38
rs773593860 Chr.8: 27598180-27598180 on Build GRCh38
rs750266639 Chr.8: 27598215-27598215 on Build GRCh38
rs899980237 Chr.8: 27598168-27598169 on Build GRCh38
rs1057085278 Chr.8: 27598353-27598353 on Build GRCh38
rs1035176289 Chr.8: 27598315-27598315 on Build GRCh38
rs773516067 Chr.8: 27598268-27598269 on Build GRCh38
rs763397906 Chr.8: 27598187-27598187 on Build GRCh38
rs776344675 Chr.8: 27598170-27598170 on Build GRCh38
rs910162613 Chr.8: 27598349-27598349 on Build GRCh38
rs964287122 Chr.8: 27598186-27598186 on Build GRCh38
rs1039833032 Chr.8: 27598154-27598154 on Build GRCh38
rs536669660 Chr.8: 27598214-27598214 on Build GRCh38
rs376490655 Chr.8: 27598190-27598190 on Build GRCh38
rs370765284 Chr.8: 27598332-27598332 on Build GRCh38
rs747765954 Chr.8: 27598174-27598174 on Build GRCh38
rs912943172 Chr.8: 27598308-27598308 on Build GRCh38
rs747280736 Chr.8: 27598269-27598285 on Build GRCh38
rs747793563 Chr.8: 27598269-27598269 on Build GRCh38
rs770127582 Chr.8: 27598178-27598178 on Build GRCh38
rs779665566 Chr.8: 27598167-27598167 on Build GRCh38
rs1050588 Chr.8: 27598158-27598158 on Build GRCh38
rs1010799843 Chr.8: 27598218-27598218 on Build GRCh38
rs754501169 Chr.8: 27598243-27598243 on Build GRCh38
rs780569687 Chr.8: 27598248-27598248 on Build GRCh38
rs113326468 Chr.8: 27598252-27598252 on Build GRCh38

View Full Product Details