Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151827372-151827621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADAC
Entrez Gene ID
13
Gene Name
arylacetamide deacetylase
Gene Aliases
CES5A1, DAC
UniGene
Hs.506908
Gene Chromosome Location
Chr.3: 151814008-151828488 on Build GRCh38
Transcripts
NM_001086.2 XM_005247104.4 NR_110202.1 NR_110203.1

Gene Symbol
AADACL2-AS1
Entrez Gene ID
101928142
Gene Name
AADACL2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.680207
Gene Chromosome Location
Chr.3: 151751179-151928175 on Build GRCh38
Transcripts
NM_001086.2 XM_005247104.4 NR_110202.1 NR_110203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3588884 c.607C>T p.L203F Chr.3: 151827579-151827579 on Build GRCh38
3588885 c.624C>T p.V208V Chr.3: 151827596-151827596 on Build GRCh38
6589204 c.630C>A p.I210I Chr.3: 151827602-151827602 on Build GRCh38
5774145 c.604-1G>C p.? Chr.3: 151827575-151827575 on Build GRCh38
479594 c.604C>G p.L202V Chr.3: 151827576-151827576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039066566 Chr.3: 151827517-151827517 on Build GRCh38
rs1015190336 Chr.3: 151827475-151827475 on Build GRCh38
rs10579932 Chr.3: 151827449-151827450 on Build GRCh38
rs572073561 Chr.3: 151827509-151827509 on Build GRCh38
rs755102871 Chr.3: 151827581-151827581 on Build GRCh38
rs554066753 Chr.3: 151827497-151827497 on Build GRCh38
rs372408554 Chr.3: 151827591-151827591 on Build GRCh38
rs913150229 Chr.3: 151827490-151827490 on Build GRCh38
rs62272918 Chr.3: 151827535-151827535 on Build GRCh38
rs369039148 Chr.3: 151827578-151827578 on Build GRCh38
rs775196735 Chr.3: 151827531-151827531 on Build GRCh38
rs1004161753 Chr.3: 151827446-151827446 on Build GRCh38
rs576256218 Chr.3: 151827512-151827512 on Build GRCh38
rs747136962 Chr.3: 151827572-151827572 on Build GRCh38
rs763445842 Chr.3: 151827537-151827537 on Build GRCh38
rs202018562 Chr.3: 151827530-151827530 on Build GRCh38
rs2410836 Chr.3: 151827534-151827534 on Build GRCh38
rs748717673 Chr.3: 151827592-151827592 on Build GRCh38
rs376418498 Chr.3: 151827469-151827472 on Build GRCh38
rs753836132 Chr.3: 151827541-151827541 on Build GRCh38
rs368082667 Chr.3: 151827534-151827535 on Build GRCh38
rs879036667 Chr.3: 151827515-151827515 on Build GRCh38
rs79797069 Chr.3: 151827571-151827571 on Build GRCh38
rs774657286 Chr.3: 151827533-151827533 on Build GRCh38
rs200378930 Chr.3: 151827448-151827449 on Build GRCh38
rs781080711 Chr.3: 151827588-151827588 on Build GRCh38
rs1043267367 Chr.3: 151827493-151827493 on Build GRCh38
rs911921175 Chr.3: 151827587-151827587 on Build GRCh38
rs992666533 Chr.3: 151827468-151827469 on Build GRCh38
rs201095759 Chr.3: 151827560-151827560 on Build GRCh38
rs11306821 Chr.3: 151827534-151827534 on Build GRCh38
rs386667110 Chr.3: 151827530-151827535 on Build GRCh38
rs759299218 Chr.3: 151827525-151827525 on Build GRCh38
rs986142347 Chr.3: 151827583-151827583 on Build GRCh38
rs760209965 Chr.3: 151827498-151827501 on Build GRCh38
rs772042255 Chr.3: 151827535-151827536 on Build GRCh38
rs2410837 Chr.3: 151827530-151827530 on Build GRCh38
rs200951194 Chr.3: 151827533-151827533 on Build GRCh38
rs945934332 Chr.3: 151827492-151827492 on Build GRCh38
rs869183137 Chr.3: 151827537-151827537 on Build GRCh38
rs180907890 Chr.3: 151827562-151827562 on Build GRCh38
rs796375851 Chr.3: 151827448-151827449 on Build GRCh38
rs765058070 Chr.3: 151827548-151827548 on Build GRCh38

View Full Product Details