Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5562789-5563018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVC2
Entrez Gene ID
132884
Gene Name
EvC ciliary complex subunit 2
Gene Aliases
LBN, WAD
UniGene
Hs.87306
Gene Chromosome Location
Chr.4: 5531921-5709548 on Build GRCh38
Transcripts
NM_001166136.1 NM_147127.4 XM_017007735.1 XM_017007736.1 XM_017007737.1 XM_017007739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6041997 c.3885T>C p.F1295F Chr.4: 5562890-5562890 on Build GRCh38
6426310 c.3869G>T p.R1290M Chr.4: 5562906-5562906 on Build GRCh38
3917930 c.3846G>C p.E1282D Chr.4: 5562929-5562929 on Build GRCh38
734161 c.3910G>A p.A1304T Chr.4: 5562865-5562865 on Build GRCh38
7156128 c.3846G>A p.E1282E Chr.4: 5562929-5562929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780751172 Chr.4: 5562906-5562906 on Build GRCh38
rs759364116 Chr.4: 5562929-5562929 on Build GRCh38
rs928695995 Chr.4: 5562820-5562820 on Build GRCh38
rs750789655 Chr.4: 5562962-5562962 on Build GRCh38
rs373925106 Chr.4: 5562892-5562892 on Build GRCh38
rs1011877594 Chr.4: 5562997-5562997 on Build GRCh38
rs764009438 Chr.4: 5562942-5562942 on Build GRCh38
rs772566880 Chr.4: 5562950-5562950 on Build GRCh38
rs765051130 Chr.4: 5562934-5562934 on Build GRCh38
rs371771308 Chr.4: 5562926-5562926 on Build GRCh38
rs781685362 Chr.4: 5562876-5562876 on Build GRCh38
rs368103321 Chr.4: 5562917-5562917 on Build GRCh38
rs587776568 Chr.4: 5562982-5562982 on Build GRCh38
rs200844130 Chr.4: 5562856-5562856 on Build GRCh38
rs764335369 Chr.4: 5562997-5562998 on Build GRCh38
rs867446852 Chr.4: 5562999-5562999 on Build GRCh38
rs770574777 Chr.4: 5562838-5562838 on Build GRCh38
rs1015050543 Chr.4: 5562853-5562853 on Build GRCh38
rs1056197547 Chr.4: 5562995-5562995 on Build GRCh38
rs966509995 Chr.4: 5562884-5562884 on Build GRCh38
rs730882232 Chr.4: 5562881-5562882 on Build GRCh38
rs921952317 Chr.4: 5562893-5562893 on Build GRCh38
rs770487741 Chr.4: 5562826-5562826 on Build GRCh38
rs765207079 Chr.4: 5562852-5562852 on Build GRCh38
rs202036951 Chr.4: 5562932-5562932 on Build GRCh38
rs771435248 Chr.4: 5562916-5562916 on Build GRCh38
rs146269526 Chr.4: 5562870-5562870 on Build GRCh38
rs780591966 Chr.4: 5562982-5562982 on Build GRCh38
rs756510211 Chr.4: 5562972-5562972 on Build GRCh38
rs760744396 Chr.4: 5562995-5562996 on Build GRCh38
rs751725959 Chr.4: 5562948-5562948 on Build GRCh38
rs752839237 Chr.4: 5562894-5562896 on Build GRCh38
rs750877993 Chr.4: 5562881-5562881 on Build GRCh38
rs776417916 Chr.4: 5562839-5562839 on Build GRCh38
rs762957778 Chr.4: 5562940-5562940 on Build GRCh38
rs923736115 Chr.4: 5562990-5562990 on Build GRCh38
rs902225088 Chr.4: 5562993-5562993 on Build GRCh38
rs202102149 Chr.4: 5562830-5562830 on Build GRCh38
rs776386860 Chr.4: 5562928-5562928 on Build GRCh38
rs772703728 Chr.4: 5562831-5562831 on Build GRCh38
rs138166440 Chr.4: 5562911-5562911 on Build GRCh38
rs751671346 Chr.4: 5562862-5562862 on Build GRCh38
rs200157468 Chr.4: 5562833-5562833 on Build GRCh38
rs894829697 Chr.4: 5562996-5562996 on Build GRCh38
rs761924204 Chr.4: 5562959-5562959 on Build GRCh38
rs746578489 Chr.4: 5562921-5562921 on Build GRCh38
rs547295889 Chr.4: 5562829-5562829 on Build GRCh38
rs1008781056 Chr.4: 5562965-5562965 on Build GRCh38
rs754241937 Chr.4: 5562991-5562991 on Build GRCh38
rs201651890 Chr.4: 5562938-5562938 on Build GRCh38
rs759466304 Chr.4: 5562844-5562844 on Build GRCh38
rs767904125 Chr.4: 5562961-5562961 on Build GRCh38
rs953355815 Chr.4: 5562908-5562908 on Build GRCh38

View Full Product Details