Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114074837-114075102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMBP
Entrez Gene ID
259
Gene Name
alpha-1-microglobulin/bikunin precursor
Gene Aliases
A1M, EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTI
UniGene
Hs.436911
Gene Chromosome Location
Chr.9: 114060127-114078472 on Build GRCh38
Transcripts
NM_001633.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3653301 c.156A>T p.K52N Chr.9: 114074964-114074964 on Build GRCh38
6598997 c.261-1G>T p.? Chr.9: 114075037-114075037 on Build GRCh38
3670403 c.159C>A p.S53S Chr.9: 114074961-114074961 on Build GRCh38
5467572 c.279G>A p.E93E Chr.9: 114075018-114075018 on Build GRCh38
3653300 c.333A>T p.K111N Chr.9: 114074964-114074964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777385730 Chr.9: 114074986-114074986 on Build GRCh38
rs751458591 Chr.9: 114074946-114074946 on Build GRCh38
rs566733427 Chr.9: 114074886-114074886 on Build GRCh38
rs138118551 Chr.9: 114075028-114075028 on Build GRCh38
rs747307471 Chr.9: 114074994-114074994 on Build GRCh38
rs552277894 Chr.9: 114074917-114074917 on Build GRCh38
rs1016607170 Chr.9: 114075061-114075061 on Build GRCh38
rs769729448 Chr.9: 114075020-114075020 on Build GRCh38
rs977820709 Chr.9: 114074941-114074941 on Build GRCh38
rs79644634 Chr.9: 114075075-114075075 on Build GRCh38
rs774714551 Chr.9: 114075062-114075062 on Build GRCh38
rs113926878 Chr.9: 114075039-114075039 on Build GRCh38
rs113351600 Chr.9: 114074959-114074959 on Build GRCh38
rs766774352 Chr.9: 114075053-114075053 on Build GRCh38
rs767492688 Chr.9: 114074952-114074952 on Build GRCh38
rs927545948 Chr.9: 114074872-114074872 on Build GRCh38
rs748722080 Chr.9: 114075016-114075016 on Build GRCh38
rs780397136 Chr.9: 114074993-114074993 on Build GRCh38
rs768048043 Chr.9: 114074960-114074960 on Build GRCh38
rs112052959 Chr.9: 114074958-114074958 on Build GRCh38
rs763274640 Chr.9: 114074927-114074927 on Build GRCh38
rs756964881 Chr.9: 114074991-114074991 on Build GRCh38
rs1050280696 Chr.9: 114075058-114075058 on Build GRCh38
rs746000339 Chr.9: 114074876-114074876 on Build GRCh38
rs750935801 Chr.9: 114074971-114074972 on Build GRCh38
rs767439904 Chr.9: 114075080-114075080 on Build GRCh38
rs150403120 Chr.9: 114075015-114075015 on Build GRCh38
rs888456476 Chr.9: 114075021-114075021 on Build GRCh38
rs200699822 Chr.9: 114074950-114074950 on Build GRCh38
rs752690306 Chr.9: 114074980-114074980 on Build GRCh38
rs765382231 Chr.9: 114074909-114074909 on Build GRCh38
rs532523634 Chr.9: 114074890-114074890 on Build GRCh38
rs755527367 Chr.9: 114074985-114074985 on Build GRCh38
rs769739285 Chr.9: 114074968-114074968 on Build GRCh38
rs113880962 Chr.9: 114075038-114075038 on Build GRCh38
rs763253684 Chr.9: 114075037-114075037 on Build GRCh38
rs766254179 Chr.9: 114074945-114074945 on Build GRCh38

View Full Product Details