Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172448332-172448593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf105
Entrez Gene ID
92346
Gene Name
chromosome 1 open reading frame 105
Gene Aliases
-
UniGene
Hs.517991
Gene Chromosome Location
Chr.1: 172418560-172468829 on Build GRCh38
Transcripts
NM_139240.3 XM_005245608.2 XM_005245610.3 XM_006711630.2 XM_011510152.2 XM_011510153.2 XM_011510154.2 XM_011510155.2 XM_011510156.2 XM_017002808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423404 c.121T>A p.S41T Chr.1: 172448454-172448454 on Build GRCh38
5991238 c.137C>A p.T46N Chr.1: 172448470-172448470 on Build GRCh38
4410930 c.147G>C p.K49N Chr.1: 172448480-172448480 on Build GRCh38
3478383 c.194C>T p.S65F Chr.1: 172448527-172448527 on Build GRCh38
3478384 c.196A>T p.K66* Chr.1: 172448529-172448529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756806935 Chr.1: 172448519-172448519 on Build GRCh38
rs757975050 Chr.1: 172448511-172448511 on Build GRCh38
rs541966045 Chr.1: 172448376-172448376 on Build GRCh38
rs753376117 Chr.1: 172448388-172448388 on Build GRCh38
rs376324202 Chr.1: 172448518-172448518 on Build GRCh38
rs372341692 Chr.1: 172448478-172448478 on Build GRCh38
rs766969605 Chr.1: 172448503-172448503 on Build GRCh38
rs554416370 Chr.1: 172448483-172448483 on Build GRCh38
rs941245862 Chr.1: 172448524-172448524 on Build GRCh38
rs774494773 Chr.1: 172448459-172448459 on Build GRCh38
rs867889558 Chr.1: 172448497-172448497 on Build GRCh38
rs530686155 Chr.1: 172448400-172448400 on Build GRCh38
rs910808687 Chr.1: 172448387-172448387 on Build GRCh38
rs144143981 Chr.1: 172448510-172448510 on Build GRCh38
rs761885038 Chr.1: 172448468-172448468 on Build GRCh38
rs150462196 Chr.1: 172448408-172448408 on Build GRCh38
rs888979713 Chr.1: 172448450-172448450 on Build GRCh38
rs150713000 Chr.1: 172448488-172448488 on Build GRCh38
rs779199899 Chr.1: 172448421-172448421 on Build GRCh38
rs368460288 Chr.1: 172448410-172448410 on Build GRCh38
rs16844474 Chr.1: 172448418-172448418 on Build GRCh38
rs763628149 Chr.1: 172448514-172448514 on Build GRCh38
rs112854516 Chr.1: 172448420-172448420 on Build GRCh38
rs533944254 Chr.1: 172448537-172448537 on Build GRCh38
rs567840448 Chr.1: 172448546-172448546 on Build GRCh38
rs768793316 Chr.1: 172448458-172448458 on Build GRCh38
rs780788649 Chr.1: 172448403-172448403 on Build GRCh38
rs568824511 Chr.1: 172448374-172448374 on Build GRCh38
rs745630889 Chr.1: 172448542-172448542 on Build GRCh38
rs761140741 Chr.1: 172448502-172448502 on Build GRCh38
rs763010358 Chr.1: 172448436-172448436 on Build GRCh38
rs755963684 Chr.1: 172448556-172448556 on Build GRCh38
rs901159416 Chr.1: 172448552-172448552 on Build GRCh38
rs563255487 Chr.1: 172448380-172448380 on Build GRCh38
rs945540387 Chr.1: 172448417-172448417 on Build GRCh38

View Full Product Details