Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34855750-34856007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM12
Entrez Gene ID
113444
Gene Name
small integral membrane protein 12
Gene Aliases
C1orf212
UniGene
Hs.27160
Gene Chromosome Location
Chr.1: 34850362-34859820 on Build GRCh38
Transcripts
NM_001164824.1 NM_001164825.1 NM_001320261.1 NM_138428.5 XM_005270403.4 XM_011540553.2 XM_011540554.2 XM_011540555.2 XM_017000166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4597143 c.157C>T p.R53W Chr.1: 34855821-34855821 on Build GRCh38
4532479 c.188G>A p.G63D Chr.1: 34855790-34855790 on Build GRCh38
5491074 c.21C>T p.T7T Chr.1: 34855957-34855957 on Build GRCh38
3805024 c.184C>G p.L62V Chr.1: 34855794-34855794 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898164579 Chr.1: 34855773-34855773 on Build GRCh38
rs759754866 Chr.1: 34855778-34855778 on Build GRCh38
rs991824937 Chr.1: 34855944-34855944 on Build GRCh38
rs753079588 Chr.1: 34855791-34855791 on Build GRCh38
rs943398951 Chr.1: 34855829-34855829 on Build GRCh38
rs933786081 Chr.1: 34855906-34855906 on Build GRCh38
rs868520467 Chr.1: 34855824-34855824 on Build GRCh38
rs764683292 Chr.1: 34855812-34855812 on Build GRCh38
rs768384287 Chr.1: 34855912-34855912 on Build GRCh38
rs535678870 Chr.1: 34855946-34855946 on Build GRCh38
rs371366501 Chr.1: 34855916-34855916 on Build GRCh38
rs751549531 Chr.1: 34855957-34855957 on Build GRCh38
rs758041671 Chr.1: 34855892-34855892 on Build GRCh38
rs914465158 Chr.1: 34855959-34855959 on Build GRCh38
rs1035932082 Chr.1: 34855869-34855869 on Build GRCh38
rs774468275 Chr.1: 34855777-34855777 on Build GRCh38
rs12733646 Chr.1: 34855982-34855982 on Build GRCh38
rs746796830 Chr.1: 34855911-34855911 on Build GRCh38
rs756751008 Chr.1: 34855795-34855795 on Build GRCh38
rs757344275 Chr.1: 34855851-34855851 on Build GRCh38
rs968032420 Chr.1: 34855974-34855974 on Build GRCh38
rs781131184 Chr.1: 34855926-34855926 on Build GRCh38
rs768043140 Chr.1: 34855785-34855785 on Build GRCh38
rs568656036 Chr.1: 34855863-34855863 on Build GRCh38
rs908404577 Chr.1: 34855950-34855950 on Build GRCh38
rs745524688 Chr.1: 34855861-34855861 on Build GRCh38
rs774468427 Chr.1: 34855987-34855987 on Build GRCh38
rs547310061 Chr.1: 34855852-34855852 on Build GRCh38
rs779894066 Chr.1: 34855901-34855901 on Build GRCh38
rs754336631 Chr.1: 34855819-34855819 on Build GRCh38
rs951097785 Chr.1: 34855804-34855804 on Build GRCh38
rs770159091 Chr.1: 34855822-34855823 on Build GRCh38

View Full Product Details