Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1090318-1090565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf159
Entrez Gene ID
54991
Gene Name
chromosome 1 open reading frame 159
Gene Aliases
-
UniGene
Hs.235095
Gene Chromosome Location
Chr.1: 1081811-1116356 on Build GRCh38
Transcripts
NM_017891.4 XM_005244765.2 XM_006710717.2 XM_006710718.2 XM_011541651.2 XM_011541652.2 XM_011541653.2 XM_011541655.2 XM_011541656.2 XM_011541658.2 XM_011541659.2 XM_011541660.2 XM_011541661.2 XM_011541662.2 XM_011541663.2 XM_011541665.2 XM_017001572.1 XM_017001573.1 XM_017001574.1 XM_017001575.1 XM_017001576.1 XM_017001577.1 XM_017001578.1 XM_017001579.1 XM_017001580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196800 c.109G>A p.V37I Chr.1: 1090392-1090392 on Build GRCh38
1953715 c.108C>T p.G36G Chr.1: 1090393-1090393 on Build GRCh38
5429401 c.193G>A p.E65K Chr.1: 1090416-1090416 on Build GRCh38
6338139 c.140G>T p.C47F Chr.1: 1090361-1090361 on Build GRCh38
6338140 c.248G>T p.C83F Chr.1: 1090361-1090361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774604062 Chr.1: 1090339-1090339 on Build GRCh38
rs553118152 Chr.1: 1090417-1090417 on Build GRCh38
rs776275320 Chr.1: 1090463-1090463 on Build GRCh38
rs752556018 Chr.1: 1090541-1090541 on Build GRCh38
rs771806426 Chr.1: 1090459-1090459 on Build GRCh38
rs370337124 Chr.1: 1090393-1090393 on Build GRCh38
rs530807127 Chr.1: 1090510-1090510 on Build GRCh38
rs544082255 Chr.1: 1090460-1090460 on Build GRCh38
rs928684394 Chr.1: 1090502-1090503 on Build GRCh38
rs145234046 Chr.1: 1090416-1090416 on Build GRCh38
rs191506077 Chr.1: 1090356-1090356 on Build GRCh38
rs948782092 Chr.1: 1090341-1090341 on Build GRCh38
rs907619644 Chr.1: 1090350-1090350 on Build GRCh38
rs368019045 Chr.1: 1090441-1090441 on Build GRCh38
rs371260854 Chr.1: 1090418-1090418 on Build GRCh38
rs202040915 Chr.1: 1090386-1090386 on Build GRCh38
rs866575703 Chr.1: 1090464-1090464 on Build GRCh38
rs557030242 Chr.1: 1090372-1090372 on Build GRCh38
rs575483653 Chr.1: 1090443-1090443 on Build GRCh38
rs970399497 Chr.1: 1090473-1090473 on Build GRCh38
rs891316563 Chr.1: 1090396-1090396 on Build GRCh38
rs372044064 Chr.1: 1090462-1090462 on Build GRCh38
rs1029332512 Chr.1: 1090540-1090540 on Build GRCh38
rs370717212 Chr.1: 1090512-1090512 on Build GRCh38
rs376055242 Chr.1: 1090392-1090392 on Build GRCh38
rs779444758 Chr.1: 1090492-1090492 on Build GRCh38
rs758539901 Chr.1: 1090511-1090511 on Build GRCh38
rs544923479 Chr.1: 1090435-1090435 on Build GRCh38
rs759949963 Chr.1: 1090467-1090467 on Build GRCh38
rs377138852 Chr.1: 1090532-1090532 on Build GRCh38
rs766857900 Chr.1: 1090363-1090363 on Build GRCh38
rs753032271 Chr.1: 1090477-1090477 on Build GRCh38
rs996509419 Chr.1: 1090539-1090539 on Build GRCh38
rs532633963 Chr.1: 1090497-1090497 on Build GRCh38
rs28469423 Chr.1: 1090479-1090479 on Build GRCh38
rs569418339 Chr.1: 1090508-1090508 on Build GRCh38
rs552533735 Chr.1: 1090504-1090504 on Build GRCh38
rs929416715 Chr.1: 1090507-1090507 on Build GRCh38
rs374160739 Chr.1: 1090472-1090472 on Build GRCh38
rs755400332 Chr.1: 1090482-1090482 on Build GRCh38
rs565080850 Chr.1: 1090442-1090442 on Build GRCh38
rs186537688 Chr.1: 1090345-1090345 on Build GRCh38
rs3813203 Chr.1: 1090481-1090481 on Build GRCh38
rs754309882 Chr.1: 1090364-1090364 on Build GRCh38
rs78662953 Chr.1: 1090528-1090528 on Build GRCh38
rs760202921 Chr.1: 1090461-1090461 on Build GRCh38
rs1031846483 Chr.1: 1090475-1090477 on Build GRCh38
rs75079403 Chr.1: 1090371-1090371 on Build GRCh38
rs549405365 Chr.1: 1090533-1090533 on Build GRCh38

View Full Product Details